Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- CAS No.:30827-99-7
9087-70-1
66701-25-5
103476-89-7 - Description:Protease inhibitor cocktail, EDTA free
- Size:20 tablet
- Storage Conditions:−20 °C
- Appearance:White to off-white tablets
- Cat. No.:77510-410
Protease inhibitor cocktail, EDTA-free tablets are designed to protect isolated proteins by providing an extraordinary inhibition of serine proteases, cysteine proteases, and aminopeptidases during extractions from animal and plant tissues or cells, yeast, and bacteria. The provided tablets do not contain EDTA, leaving the stability and the function of metal-depended proteins unaffected. Contains both irreversible and reversible protease inhibitors. Aspartic and metallo proteases are not inhibited with the exception of aminopeptidases. The affinity purification of Poly-His tagged fusion proteins via IMAC (immobilized metal ion affinity chromatography) is also facilitated (no dialysis necessary). Tablets contain AEBSF, Aprotinin, E-64, and Leupeptin hemisulfate.
Preparation: Dissolve tablets directly in the extraction medium. Depending on the proteolytic activity, one tablet should be sufficient for the inhibition of the existing proteolytic activity in 50 ml. If very high proteolytic activity is present, one tablet should be used for 25 ml extraction buffer.