Superfrost® Clipped Corner Slides, Erie Scientific (Thermo Scientific)
Catalog # 48404-472
Supplier: Erie Scientific
Specifications
- Description:Slides, Superfrost
- L×W:75×25 mm (3×1")
- Thickness:1.0 mm
- Environmentally Preferable:
- Cat. No.:48404-472
- Supplier no.:4951WS44370
Specifications
About this item
Ideal for technicians who routinely prepare large numbers of slides, these 45° clipped corner slides offer exceptional cleanliness and resistance to sticking
This design allows for safer handling, as corners are less likely to chip and specimens are drawn away from the edges. The durable, opaque printed surfaces are inert to common laboratory chemicals and reagents. Designed specifically for use with Sysmex® SP-1, SP-100, and SP-1000 instruments. Slides are autoclavable.