PCR Filming Scraper
Catalog # 77508-078
Supplier: NEST SCIENTIFIC
Specifications
- Color:Blue
- Description:PCR filming scraper
- Dimensions:70×78 mm
- Material:TPE
- Cat. No.:77508-078
Specifications
About this item
Using with sealing film to make the sealing film fit better with the PCR plate.