Hypersil™ Chiral HPLC Columns
About this item
Separate a wide range of enantiomers with Hypersil™ Chiral HPLC columns, designed for separations of neutral, acidic and basic racemates in normal phase. These chiral columns can help ensure your purification needs are met by resolving racemic mixtures and potential contaminants so you can be confident in the purity of target compound. To maximize screening efficiency, these columns can be ordered in kits which contain a column in each of our chiral phases. All columns are available in analytical and preparative scale formats.
- Standard threaded and flange hardware for standard applications, expertly packed and robustly designed to provide versatile products for routine use
- Standard threaded hardware for column I.D. <40 mm
- Standard flange hardware for column I.D. ≥40 mm
- Standard hardware can be modified on request to be compatible with SFC
Hypersil Chiral JT: Best starting point for method development. Chemistry: Tris-(4-methylbenzoyl)-cellulose modified silica. USP L80.
Hypersil Chiral AT: Highest generality of all chiral phases, No aromatic ring necessary, No inversion of elution order possible. Chemistry: Amylose tris-3,5-dimethylphenylcarbamate modified silica. USP L51.
Hypersil Chiral OT: Highly versatile chiral phases. No aromatic ring necessary. No inversion of elution order possible. Chemistry: Cellulose tris-(3.5-dimethylphenyl-carbamate) modified silica. USP L40.
39 Options Available Below
Product Details & Documents
Separate a wide range of enantiomers with Hypersil™ Chiral HPLC columns, designed for separations of neutral, acidic and basic racemates in normal phase. These chiral columns can help ensure your purification needs are met by resolving racemic mixtures and potential contaminants so you can be confident in the purity of target compound. To maximize screening efficiency, these columns can be ordered in kits which contain a column in each of our chiral phases. All columns are available in analytical and preparative scale formats.
- Standard threaded and flange hardware for standard applications, expertly packed and robustly designed to provide versatile products for routine use
- Standard threaded hardware for column I.D. <40 mm
- Standard flange hardware for column I.D. ≥40 mm
- Standard hardware can be modified on request to be compatible with SFC
Hypersil Chiral JT: Best starting point for method development. Chemistry: Tris-(4-methylbenzoyl)-cellulose modified silica. USP L80.
Hypersil Chiral AT: Highest generality of all chiral phases, No aromatic ring necessary, No inversion of elution order possible. Chemistry: Amylose tris-3,5-dimethylphenylcarbamate modified silica. USP L51.
Hypersil Chiral OT: Highly versatile chiral phases. No aromatic ring necessary. No inversion of elution order possible. Chemistry: Cellulose tris-(3.5-dimethylphenyl-carbamate) modified silica. USP L40.
Specifications for Product Family
Specifications
- Catalog No:
- 76331-924
- 76331-926
- 76331-928
- 76331-930
- 76331-932
- 76331-934
- 76331-936
- 76331-938
- 76331-940
- 76331-942
- 76331-944
- 76331-896
- 76331-898
- 76331-900
- 76331-902
- 76331-904
- 76331-906
- 76331-908
- 76331-910
- 76331-912
- 76331-914
- 76331-916
- 76331-918
- 76331-920
- 76331-922
- 76331-868
- 76331-870
- 76331-872
- 76331-874
- 76331-876
- 76331-878
- 76331-880
- 76331-882
- 76331-884
- 76331-886
- 76331-888
- 76331-890
- 76331-892
- 76331-894
- Max. Pressure:
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- 255 bar (3700 psi)
- Surface Area:
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
- 30 m²/g
Specifications
Product Family Options
Hypersil Chiral JT columns
Product Information
- Column FormatPore sizeParticle SizeØ int.LengthPacking materialAvailabilityPrice
- Analytical1000 Å3 µm4.6 mm100 mmSpherical, fully porous silicaLog in for availabilityEach$2,004.34Supplier Number: 42203-104630Specifications:
More
Specifications
Cat. No.76331-924Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Analytical1000 Å3 µm4.6 mm150 mmSpherical, fully porous silicaLog in for availabilityEach$2,327.70Supplier Number: 42203-154630Specifications:
More
Specifications
Cat. No.76331-926Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Analytical1000 Å3 µm4.6 mm250 mmSpherical, fully porous silicaLog in for availabilityEach$3,305.13Supplier Number: 42203-254630Specifications:
More
Specifications
Cat. No.76331-928Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Analytical1000 Å5 µm4.6 mm100 mmSpherical, fully porous silicaLog in for availabilityEach$1,786.74Supplier Number: 42205-104630Specifications:
More
Specifications
Cat. No.76331-930Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Analytical1000 Å5 µm4.6 mm150 mmSpherical, fully porous silicaLog in for availabilityEach$2,004.34Supplier Number: 42205-154630Specifications:
More
Specifications
Cat. No.76331-932Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Analytical1000 Å5 µm4.6 mm250 mmSpherical, fully porous silicaLog in for availabilityEach$2,327.70Supplier Number: 42205-254630Specifications:
More
Specifications
Cat. No.76331-934Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Analytical1000 Å10 µm4.6 mm100 mmSpherical, fully porous silicaLog in for availabilityEach$1,778.50Supplier Number: 42210-104630Specifications:
More
Specifications
Cat. No.76331-936Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Analytical1000 Å10 µm4.6 mm150 mmSpherical, fully porous silicaLog in for availabilityEach$1,995.10Supplier Number: 42210-154630Specifications:
More
Specifications
Cat. No.76331-938Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Analytical1000 Å10 µm4.6 mm250 mmSpherical, fully porous silicaLog in for availabilityEach$2,327.70Supplier Number: 42210-254630Specifications:
More
Specifications
Cat. No.76331-940Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Preparative1000 Å5 µm10 mm250 mmSpherical, fully porous silicaLog in for availabilityEach$8,398.01Supplier Number: 42205-259070ASpecifications:
More
Specifications
Cat. No.76331-942Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Preparative1000 Å10 µm10 mm250 mmSpherical, fully porous silicaLog in for availabilityEach$7,258.27Supplier Number: 42210-259070ASpecifications:
More
Specifications
Cat. No.76331-944Max. Pressure255 bar (3700 psi)Surface Area30 m²/g
Hypersil Chiral AT columns
Product Information
- Column FormatPore sizeParticle SizeØ int.LengthPacking materialAvailabilityPrice
- Supplier Number: 42103-054630Specifications:
More
Specifications
Cat. No.76331-896Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Analytical1000 Å3 µm4.6 mm100 mmSpherical, fully porous silicaLog in for availabilityEach$1,762.20Supplier Number: 42103-104630Specifications:
More
Specifications
Cat. No.76331-898Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Analytical1000 Å3 µm4.6 mm150 mmSpherical, fully porous silicaLog in for availabilityEach$2,306.83Supplier Number: 42103-154630Specifications:
More
Specifications
Cat. No.76331-900Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Analytical1000 Å3 µm4.6 mm250 mmSpherical, fully porous silicaLog in for availabilityEach$3,258.68Supplier Number: 42103-254630Specifications:
More
Specifications
Cat. No.76331-902Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Analytical1000 Å5 µm4.6 mm100 mmSpherical, fully porous silicaLog in for availabilityEach$1,770.36Supplier Number: 42105-104630Specifications:
More
Specifications
Cat. No.76331-904Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Analytical1000 Å5 µm4.6 mm150 mmSpherical, fully porous silicaLog in for availabilityEach$1,983.46Supplier Number: 42105-154630Specifications:
More
Specifications
Cat. No.76331-906Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Analytical1000 Å5 µm4.6 mm250 mmSpherical, fully porous silicaLog in for availabilityEach$2,306.83Supplier Number: 42105-254630Specifications:
More
Specifications
Cat. No.76331-908Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Analytical1000 Å10 µm4.6 mm100 mmSpherical, fully porous silicaLog in for availabilityEach$1,762.20Supplier Number: 42110-104630Specifications:
More
Specifications
Cat. No.76331-910Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Analytical1000 Å10 µm4.6 mm150 mmSpherical, fully porous silicaLog in for availabilityEach$1,974.32Supplier Number: 42110-154630Specifications:
More
Specifications
Cat. No.76331-912Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Analytical1000 Å10 µm4.6 mm250 mmSpherical, fully porous silicaLog in for availabilityEach$2,306.83Supplier Number: 42110-254630Specifications:
More
Specifications
Cat. No.76331-914Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Preparative1000 Å5 µm10 mm250 mmSpherical, fully porous silicaLog in for availabilityEach$8,398.01Supplier Number: 42105-259070ASpecifications:
More
Specifications
Cat. No.76331-916Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Preparative1000 Å5 µm20 mm250 mmSpherical, fully porous silicaLog in for availabilityEach$17,079.31Supplier Number: 42105-259270ASpecifications:
More
Specifications
Cat. No.76331-918Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Preparative1000 Å10 µm10 mm250 mmSpherical, fully porous silicaLog in for availabilityEach$6,700.92Supplier Number: 42110-259070ASpecifications:
More
Specifications
Cat. No.76331-920Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Preparative1000 Å10 µm20 mm250 mmSpherical, fully porous silicaLog in for availabilityEach$14,452.78Supplier Number: 42110-259270ASpecifications:
More
Specifications
Cat. No.76331-922Max. Pressure255 bar (3700 psi)Surface Area30 m²/g
Hypersil Chiral OT columns
Product Information
- Column FormatPore sizeParticle SizeØ int.LengthPacking materialAvailabilityPrice
- Supplier Number: 42003-054630Specifications:
More
Specifications
Cat. No.76331-868Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Analytical1000 Å3 µm4.6 mm100 mmSpherical, fully porous silicaLog in for availabilityEach$1,770.36Supplier Number: 42003-104630Specifications:
More
Specifications
Cat. No.76331-870Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Analytical1000 Å3 µm4.6 mm150 mmSpherical, fully porous silicaLog in for availabilityEach$2,306.83Supplier Number: 42003-154630Specifications:
More
Specifications
Cat. No.76331-872Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Analytical1000 Å3 µm4.6 mm250 mmSpherical, fully porous silicaLog in for availabilityEach$3,273.80Supplier Number: 42003-254630Specifications:
More
Specifications
Cat. No.76331-874Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Analytical1000 Å5 µm4.6 mm100 mmSpherical, fully porous silicaLog in for availabilityEach$1,770.36Supplier Number: 42005-104630Specifications:
More
Specifications
Cat. No.76331-876Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Analytical1000 Å5 µm4.6 mm150 mmSpherical, fully porous silicaLog in for availabilityEach$1,974.32Supplier Number: 42005-154630Specifications:
More
Specifications
Cat. No.76331-878Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Analytical1000 Å5 µm4.6 mm250 mmSpherical, fully porous silicaLog in for availabilityEach$2,306.83Supplier Number: 42005-254630Specifications:
More
Specifications
Cat. No.76331-880Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Analytical1000 Å10 µm4.6 mm100 mmSpherical, fully porous silicaLog in for availabilityEach$1,762.20Supplier Number: 42010-104630Specifications:
More
Specifications
Cat. No.76331-882Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Analytical1000 Å10 µm4.6 mm150 mmSpherical, fully porous silicaLog in for availabilityEach$1,974.32Supplier Number: 42010-154630Specifications:
More
Specifications
Cat. No.76331-884Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Analytical1000 Å10 µm4.6 mm250 mmSpherical, fully porous silicaLog in for availabilityEach$2,296.17Supplier Number: 42010-254630Specifications:
More
Specifications
Cat. No.76331-886Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Preparative1000 Å5 µm10 mm250 mmSpherical, fully porous silicaLog in for availabilityEach$8,398.01Supplier Number: 42005-259070ASpecifications:
More
Specifications
Cat. No.76331-888Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Preparative1000 Å5 µm20 mm250 mmSpherical, fully porous silicaLog in for availabilityEach$17,044.78Supplier Number: 42005-259270ASpecifications:
More
Specifications
Cat. No.76331-890Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Preparative1000 Å10 µm10 mm250 mmSpherical, fully porous silicaLog in for availabilityEach$6,700.92Supplier Number: 42010-259070ASpecifications:
More
Specifications
Cat. No.76331-892Max. Pressure255 bar (3700 psi)Surface Area30 m²/g - Preparative1000 Å10 µm20 mm250 mmSpherical, fully porous silicaLog in for availabilityEach$14,452.78Supplier Number: 42010-259270ASpecifications:
More
Specifications
Cat. No.76331-894Max. Pressure255 bar (3700 psi)Surface Area30 m²/g