ContactUsLinkComponent
Turbidity Standard, 5 NTU, Polymer Bead Suspension
Turbidity Standard, 5 NTU, Polymer Bead Suspension
Catalog # RCR8805500500G
Supplier:  Ricca Chemical
CAS Number:  
undefined
Turbidity Standard, 5 NTU, Polymer Bead Suspension
Catalog # RCR8805500500G
Supplier:  Ricca Chemical
Supplier Number:  R8805500-500G
CAS Number:  
Specifications
  • Pack type:
    Plastic
  • Turbidity:
    5 NTU
  • Size:
    500 ml
  • Cat. No.:
    RCR8805500500G
Specifications
About this item
Traceable to commercially prepared EPA recognized primary standards.
  • Ready to use solution
Polymer bead suspension.