ContactUsLinkComponent
Turbidity Standard, 6 NTU, Polymer Bead Suspension
Turbidity Standard, 6 NTU, Polymer Bead Suspension
Catalog # RCR8805600100C
Supplier:  Ricca Chemical
CAS Number:  
undefined
Turbidity Standard, 6 NTU, Polymer Bead Suspension
Catalog # RCR8805600100C
Supplier:  Ricca Chemical
Supplier Number:  R8805600-100C
CAS Number:  
Specifications
  • Pack type:
    Glass
  • Turbidity:
    6 NTU
  • Size:
    100 ml
  • Cat. No.:
    RCR8805600100C
Specifications
About this item
Traceable to commercially prepared EPA recognized primary standards.
  • Ready to use solution
Polymer bead suspension.