CellPro™ M-15 Buffered Phosphate, 10X
Supplier: Alkali Scientific
Certificates
About this item
CellPro™ M-15 Buffered phosphate is meticulously formulated to meet the stringent demands of molecular biology research.
- Sterile and endotoxin tested
- Ideal for laboratory testing, biopharmaceutical R and D
- Ideal in research, testing, cell culture and molecular protocols
- Contains no antimicrobial agents
Designed with precision, this solution offers a balanced composition of essential buffering agents, ensuring optimal pH stability for a wide range of experimental conditions. Through advanced sterile filtration techniques, it maintains the integrity and purity of each batch, minimizing the risk of contamination and ensuring reproducible results. Ideal for nucleic acid isolation, enzymatic reactions, and DNA electrophoresis, CellPro™ M-15 Buffered phosphate is a cornerstone reagent for molecular biologists seeking reliability and performance in their experiments.
Caution: UN No. 3248
Specifications
- CAS No.:7558-79-4;7778-77-6;7732-18-5
Specifications
Frequently Bought Together