Iron Standard Stock, USP 241 for Iron
Catalog # RCR4199000500A
Supplier: Ricca Chemical
CAS Number:
Specifications
- Concentration:1000 ppm
- Pack type:Plastic bottle
- Description:Iron standard stock, USP
- Size:500 ml
- Cat. No.:RCR4199000500A
- Element:Iron
Specifications
About this item
Iron stock standard.
- Conforms to preparation per USP 241 for Iron
Stock solution for analysis.
Certifications: USP 241 for Iron.