Nitrate Standard, 2 ppm NO₃⁻
Catalog # RCR5307002500A
Supplier: Ricca Chemical
CAS Number:
Specifications
- Concentration:2 ppm
- Matrix:H₂O
- Pack type:Plastic bottle
- Description:Nitrate standard, 2 ppm NO₃⁻
- Size:500 ml
- Cat. No.:RCR5307002500A
- Element:Nitrate
Specifications
About this item
2 ppm NO₃ Calcium Standard for analysis.
- Single element standard
Ideal for instrument calibration.