Order Entry
United States
ContactUsLinkComponent
 
Fluoride Standard, 0.4 ppm F⁻
Catalog # RCR3165440500A
Supplier:  Ricca Chemical
CAS Number:  
undefined
Fluoride Standard, 0.4 ppm F⁻
Catalog # RCR3165440500A
Supplier:  Ricca Chemical
Supplier Number:  R3165440-500A
CAS Number:  

Specifications

  • Concentration:
    0.4 ppm
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Description:
    Fluoride standard, 0.4 ppm F⁻
  • Size:
    500 ml
  • Cat. No.:
    RCR3165440500A
  • Element:
    Fluoride

Specifications

About this item

0 4 ppm F Calcium standard for analysis.

  • Single element standard

Ideal for instrument calibration.