Calcium Standard, 500 ppm Ca
Catalog # RCR1799000500A
Supplier: Ricca Chemical
CAS Number:
Specifications
- Concentration:500 ppm
- Matrix:H₂O
- Pack type:Plastic bottle
- Description:Calcium standard, 500 ppm Ca
- Size:500 ml
- Cat. No.:RCR1799000500A
- Element:Calcium
Specifications
About this item
500 ppm Calcium standard for analysis.
- Single element standard
Ideal for instrument calibration.