Order Entry
United States
Orders LinkContactUsLinkComponent
Centrifuge Tubes, Polypropylene

Centrifuge Tubes, Polypropylene

Supplier:   NEST SCIENTIFIC
Centrifuge Tubes, Polypropylene
Supplier:  NEST SCIENTIFIC
Certificates
Centrifuge Tubes, Polypropylene

4 Options Available Below

Product Details & Documents

The 250 and 500 ml centrifuge tubes can process a large number of samples at one time and are suitable for separation and harvest of large cells, supernatants, bacteria, yeast and tissue samples, therefore they are widely used in the field of life sciences and clinical experiments and can meet the requirements of biological analysis.

  • The unique sealing structure design of the cover provides a two-way security to guarantee seal ability
  • The thickened design of the conical bottom increases the maximum centrifugal limit
  • Maximum centrifugal limit 7000×g
  • Polypropylene tube
  • Tolerable temperature –80 to +120 ℃
  • The clear scale is convenient for data reading
  • DNase, RNase and pyrogen free, sterile
  • Electron beam sterilization, SAL= 10⁻⁶

Application: Separation of sediments after chemical reactions for harvest and application, harvest of biomacromolecules, inorganics and organics, in biochemistry and other biological fields, usually used to centrifuge and collect a large volume of cells, biomacromolecules and supernatants.

Product Family Options

Product Information

  • Capacity
    Material
    Color
    Cap included
    Temperature range
    Max. rcf
    Packaging
    Sterility
    Tube Bottom Shape
    Availability
    Price
  • 77508-418
    Centrifuge tube, sterilized
    Centrifuge tube, sterilized
    250 ml
    PP
    Clear
    Yes
    –80...+120 ℃
    7000×g
    6 pieces per pack, 102 pieces per case
    Sterile
    Conical bottom
    Log in for availability
    Log in for availability
    Case of 102
    Case of 102 - $405.30
    $405.30
    Case of 102
       $405.30

    Supplier Number: 622001
  • 77508-420
    Centrifuge tubes packed in the rack, sterilized
    Centrifuge tubes packed in the rack, sterilized
    250 ml
    PP
    Clear
    Yes
    –80...+120 ℃
    7000×g
    6 pieces per rack, 36 pieces per case
    Sterile
    Conical bottom
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $119.71
    $119.71
    Case of 24
       $119.71

    Supplier Number: 622002
  • 77508-422
    Centrifuge tube, sterilized
    Centrifuge tube, sterilized
    500 ml
    PP
    Clear
    Yes
    –80...+120 ℃
    7000×g
    6 pieces per pack, 24 pieces per case
    Sterile
    Conical bottom
    Log in for availability
    Log in for availability
    Case of 36
    Case of 36 - $193.33
    $193.33
    Case of 36
       $193.33

    Supplier Number: 623001
  • 77508-424
    Centrifuge tubes packed in the rack, sterilized
    Centrifuge tubes packed in the rack, sterilized
    500 ml
    PP
    Clear
    Yes
    –80...+120 ℃
    7000×g
    4 pieces per rack, 24 pieces per case
    Sterile
    Conical bottom
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $162.10
    $162.10
    Case of 24
       $162.10

    Supplier Number: 623002
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.