About this item
NEST automation/robotic tips can be used for research and development and other commonly used applications with needs of high-throughput pipetting in genomics, proteomics, cytomics, immunoassays, metabolomics, biopharmaceuticals.
- Adopt medical grade polypropylene material, which is manufactured by precision mold, and the material batch is stable
- DNase-and RNase-free, no heat source contamination
- Electron beam sterilization
- Manufacturing facility equipped with class 100,000 clean-room, products packing under sterile condition
- Unique process technology ensures that the inner surface of the tip is smooth and reduces liquid residue
- Online testing equipment is used to control the size and concentricity of products in the production line
- Both conductive and clear are available
Compatible with Hamilton/Tecan/Beckman/Agilent Systems. Filter or non-filter style can be selectable, it has super hydrophilicity, it can shield aerosol, and eliminate the risk of cross-contamination of samples.
Variety of packaging are available. Carry out structural drawing and measurement according to the original adapter to ensure that the product has good air tightness and adaptability.
Effective quality inspections are conducted for strict quality management and control per the client’s needs, including tests for DNase, RNase, proteases and pyrogens.
Beckman robotic tips are used in Beckman FX/NX,Biomek i5/ i7 and Biomek 3000.