Order Entry
United States
ContactUsLinkComponent
Casework, Laminate, Sitting Height Base Cabinets, Cupboard Cabinet
Casework, Laminate, Sitting Height Base Cabinets, Cupboard Cabinet
Catalog # 33501-086
Supplier:  Case Systems
Small Business Enterprise
undefined
Casework, Laminate, Sitting Height Base Cabinets, Cupboard Cabinet
Catalog # 33501-086
Supplier:  Case Systems
Supplier Number:  B0120302424
Small Business Enterprise

Specifications

  • Depth:
    24"
  • Description:
    Cupboard, 1 shelf, 1 right door
  • Height:
    29½"
  • Length:
    24"
  • Material:
    Plastic laminate
  • Environmentally Preferable:
  • Cat. No.:
    33501-086
  • Supplier no.:
    B0120302424

Specifications

About this item

Case Systems plastic laminate furniture combines modular and fixed laminate casework with coordinating hardware, trim, countertops, fixtures and many complementary accessories.

  • Color: beige
  • They are as attractive as they are durable
  • Case cabinetry that are adaptability necessary in today’s laboratories
  • The key component of casework is the plastic laminate

A perfect choice for laboratory environments, plastic laminate provides superior resistance to chemicals, scratching, and abrasion. The radiused 3 mm PVC edging on doors and drawers further protects the casework from dents and marring while eliminating sharp edges and corners.

The high-quality laboratory grade plastic laminate cabinets are manufactured to exact tolerances with consistent quality standards. Construction features an industrial-grade, environmentally responsible WOODSTALK® engineered board core from Dow BioProducts, using formaldehyde-free resin to bind wheat-straw fiber, and heavy-duty institutional grade hardware.

The unique hardware system provides rigid support for vertical integration without limiting adjustability. Vertical supports are slotted, allowing infinite height adjustment of overhead cabinets, electrical raceways, services, and reagent shelves.

Ordering information: Case Systems laminate laboratory furniture has all the components necessary for a complete laboratory set-up. For further information, contact your Avantor Furniture Specialist.