Order Entry
United States
ContactUsLinkComponent
 
Deactivated Silica Tubing
Catalog # 76311-738
Supplier:  Thermo Scientific
Deactivated Silica Tubing
Catalog # 76311-738
Supplier:  Thermo Scientific
Supplier Number:  60201-399

Specifications

  • Ø:
    150 µm
  • Ext.Ø:
    0.363 mm
  • Length:
    240 mm
  • Material:
    Deactivated silica
  • Environmentally Preferable:
  • Cat. No.:
    76311-738

Specifications

About this item

Achieve consistent results with this deactivated silica tubing for GC capillary applications.

  • Chemical inertness
  • High resistance