NovaStat™ Anti-Static Tapes, Micronova
Supplier: Micronova
Certificates
About this item
A transparent amber tape, specially designed with non-hygroscopic properties for sealing and packaging in ESD-sensitive environments.
- For sealing and packaging in ESD-sensitive environments
- Unaffected by humidity
- Available in a variety of sizes
- Wound on a 3" plastic core
The Micronova NovaStat™ Anti-Static Tape is made with cellophane backing and a rubber based adhesive. It will not create a static charge upon sealing or removal.
Color: Transparent amber
Frequently Bought Together