ContactUsLinkComponent
Advanced Indicator for Industial Applications
Advanced Indicator for Industial Applications
Catalog # 77434-816
Supplier:  Ohaus
undefined
Advanced Indicator for Industial Applications
Catalog # 77434-816
Supplier:  Ohaus
Supplier Number:  30126959
Specifications
  • Model:
    T72XW
  • Description:
    Advanced indicator for industial applications
  • Cat. No.:
    77434-816
Specifications
About this item
Advanced performance stainless steel indicator with external device control capabilities.
With a 50000 displayed resolution and the ability to support up to ten load cells, T72XW was designed for heavy-duty industrial applications such as vehicle weighing. Two option slots are available to enable a wide range of connectivity interfaces. Control external devices using the discrete I/O interface and transfer data through the serial, USB or Ethernet ports. T72XW is very easy to set up and navigate - featuring a vivid graphic display with on-screen prompts and an alphanumeric keypad with designated keys for common functions such as Tare and ID information.
The OHAUS T72XW indicator provides the flexibility, performance and operation necessary for specific applications with connectivity and data output capabilities - including vehicle weighing, over/under checkweighing, animal weighing and counting. The 7000 series indicator has multiple functions, intuitive software, multiple connectivity options, and the durability to stand up to the wear-and-tear in industrial settings.