Orders LinkContactUsLinkComponent
Vacuum Filtration Systems
Vacuum Filtration Systems
Supplier:  NEST SCIENTIFIC
Certificates
Vacuum Filtration Systems
60 Options Available Below
Product Details & Documents
Nest disposable vacuum filtration system is useful for large volume sample separation and sterilization for tissue culture media and other biological buffers by using a vacuum pump to provide differential pressure for the filtration.
  • High flow rates and flux
  • Low protein binding and low chemical extractable
  • Non-Pyrogenic, DNase/Rnase free
  • Sterilized by E-beam, SAL=10⁻⁶
  • Individually packaged in sterile bag
  • Hydrophilic
The units include PES or PVDF membrane filters with graduated filter tops made with polystyrene, and Acrylonitrile-Butadiene-Styrene necks with polystyrene receiving bottles. A separated sterile polyethylene cap is included.
Filtration system is manufactured from raw material of high transparency polystyrene (PS) in three different volumes: 250, 500 and 1000 ml. They are available in three styles: Complete filter/storage bottle systems, bottle top filters, and receiving bottles only.
PES application direction: For general culture media and aqueous solutions, polar or middlepolar solvents, neutral aqueous solutions.
PVDF application direction: May be used for aqueous solutions and most solvents, including strong non-polar solvents. Ideal for preparations of HPLC and GC.
The product has been irradiated and dosimetrically released based on ANSI/AAMI/ISO 11137.
Withstand temperature range: Maximum temperature 45 ℃
Shelf life: 3 years
 : Should not be used for chloroform, esters, amides and strong acids or strong bases.
Product Family Options
PVDF filter membranes
Product Information
  • Capacity
    Material
    Membrane
    Pore size
    Sterile
    Availability
    Price
  • 77507-922
    Vacuum filtration system
    Vacuum filtration system
    50 ml
    Polystyrene
    PVDF
    0.10 µm
    +
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $159.13
    $159.13
    Case of 12
       $159.13
    Supplier Number: 347102
  • 77507-920
    Vacuum filtration system
    Vacuum filtration system
    50 ml
    Polystyrene
    PVDF
    0.22 µm
    +
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $159.13
    $159.13
    Case of 12
       $159.13
    Supplier Number: 347101
  • 77507-924
    Vacuum filtration system
    Vacuum filtration system
    50 ml
    Polystyrene
    PVDF
    0.45 µm
    +
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $159.13
    $159.13
    Case of 12
       $159.13
    Supplier Number: 347103
  • 77507-898
    Vacuum filtration system
    Vacuum filtration system
    150 ml
    Polystyrene
    PVDF
    0.10 µm
    +
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $152.17
    $152.17
    Case of 12
       $152.17
    Supplier Number: 346102
  • 77507-896
    Vacuum filtration system
    Vacuum filtration system
    150 ml
    Polystyrene
    PVDF
    0.22 µm
    +
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $152.17
    $152.17
    Case of 12
       $152.17
    Supplier Number: 346101
  • 77507-900
    Vacuum filtration system
    Vacuum filtration system
    150 ml
    Polystyrene
    PVDF
    0.45 µm
    +
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $152.17
    $152.17
    Case of 12
       $152.17
    Supplier Number: 346103
  • 77507-812
    Vacuum filtration system
    Vacuum filtration system
    1000 ml
    Polystyrene
    PVDF
    0.10 µm
    +
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $295.12
    $295.12
    Case of 12
       $295.12
    Supplier Number: 344102
  • 77507-810
    Vacuum filtration system
    Vacuum filtration system
    1000 ml
    Polystyrene
    PVDF
    0.22 µm
    +
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $295.12
    $295.12
    Case of 12
       $295.12
    Supplier Number: 344101
  • 77507-814
    Vacuum filtration system
    Vacuum filtration system
    1000 ml
    Polystyrene
    PVDF
    0.45 µm
    +
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $295.12
    $295.12
    Case of 12
       $295.12
    Supplier Number: 344103
  • 77507-786
    Vacuum filtration system
    Vacuum filtration system
    500 ml
    Polystyrene
    PVDF
    0.10 µm
    +
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $204.90
    $204.90
    Case of 12
       $204.90
    Supplier Number: 343102
  • 77507-784
    Vacuum filtration system
    Vacuum filtration system
    500 ml
    Polystyrene
    PVDF
    0.22 µm
    +
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $204.90
    $204.90
    Case of 12
       $204.90
    Supplier Number: 343101
  • 77507-788
    Vacuum filtration system
    Vacuum filtration system
    500 ml
    Polystyrene
    PVDF
    0.45 µm
    +
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $204.90
    $204.90
    Case of 12
       $204.90
    Supplier Number: 343103
  • 77507-760
    Vacuum filtration system
    Vacuum filtration system
    250 ml
    Polystyrene
    PVDF
    0.10 µm
    +
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $157.54
    $157.54
    Case of 12
       $157.54
    Supplier Number: 342102
  • 77507-758
    Vacuum filtration system
    Vacuum filtration system
    250 ml
    Polystyrene
    PVDF
    0.22 µm
    +
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $157.54
    $157.54
    Case of 12
       $157.54
    Supplier Number: 342101
  • 77507-762
    Vacuum filtration system
    Vacuum filtration system
    250 ml
    Polystyrene
    PVDF
    0.45 µm
    +
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $157.54
    $157.54
    Case of 12
       $157.54
    Supplier Number: 342103
PES filter membranes
Product Information
  • Capacity
    Material
    Membrane
    Pore size
    Sterile
    Availability
    Price
  • 77507-910
    Vacuum filtration system
    Vacuum filtration system
    50 ml
    Polystyrene
    PES
    0.10 µm
    +
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $155.03
    $155.03
    Case of 12
       $155.03
    Supplier Number: 347002
  • 77507-908
    Vacuum filtration system
    Vacuum filtration system
    50 ml
    Polystyrene
    PES
    0.22 µm
    +
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $155.03
    $155.03
    Case of 12
       $155.03
    Supplier Number: 347001
  • 77507-912
    Vacuum filtration system
    Vacuum filtration system
    50 ml
    Polystyrene
    PES
    0.45 µm
    +
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $155.03
    $155.03
    Case of 12
       $155.03
    Supplier Number: 347003
  • 77507-884
    Vacuum filtration system
    Vacuum filtration system
    150 ml
    Polystyrene
    PES
    0.10 µm
    +
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $147.97
    $147.97
    Case of 12
       $147.97
    Supplier Number: 346002
  • 77507-882
    Vacuum filtration system
    Vacuum filtration system
    150 ml
    Polystyrene
    PES
    0.22 µm
    +
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $147.97
    $147.97
    Case of 12
       $147.97
    Supplier Number: 346001
  • 77507-886
    Vacuum filtration system
    Vacuum filtration system
    150 ml
    Polystyrene
    PES
    0.45 µm
    +
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $147.97
    $147.97
    Case of 12
       $147.97
    Supplier Number: 346003
  • 77507-798
    Vacuum filtration system
    Vacuum filtration system
    1000 ml
    Polystyrene
    PES
    0.10 µm
    +
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $291.53
    $291.53
    Case of 12
       $291.53
    Supplier Number: 344002
  • 77507-796
    Vacuum filtration system
    Vacuum filtration system
    1000 ml
    Polystyrene
    PES
    0.22 µm
    +
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $291.53
    $291.53
    Case of 12
       $291.53
    Supplier Number: 344001
  • 77507-800
    Vacuum filtration system
    Vacuum filtration system
    1000 ml
    Polystyrene
    PES
    0.45 µm
    +
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $291.53
    $291.53
    Case of 12
       $291.53
    Supplier Number: 344003
  • 77507-772
    Vacuum filtration system
    Vacuum filtration system
    500 ml
    Polystyrene
    PES
    0.10 µm
    +
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $200.70
    $200.70
    Case of 12
       $200.70
    Supplier Number: 343002
  • 77507-770
    Vacuum filtration system
    Vacuum filtration system
    500 ml
    Polystyrene
    PES
    0.22 µm
    +
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $178.74
    $178.74
    Case of 12
       $178.74
    Supplier Number: 343001
  • 77507-774
    Vacuum filtration system
    Vacuum filtration system
    500 ml
    Polystyrene
    PES
    0.45 µm
    +
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $200.70
    $200.70
    Case of 12
       $200.70
    Supplier Number: 343003
  • 77507-746
    Vacuum filtration system
    Vacuum filtration system
    250 ml
    Polystyrene
    PES
    0.10 µm
    +
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $153.31
    $153.31
    Case of 12
       $153.31
    Supplier Number: 342002
  • 77507-744
    Vacuum filtration system
    Vacuum filtration system
    250 ml
    Polystyrene
    PES
    0.22 µm
    +
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $153.31
    $153.31
    Case of 12
       $153.31
    Supplier Number: 342001
  • 77507-748
    Vacuum filtration system
    Vacuum filtration system
    250 ml
    Polystyrene
    PES
    0.45 µm
    +
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $153.31
    $153.31
    Case of 12
       $153.31
    Supplier Number: 342003
Bottle top vacuum filter only
Product Information
  • Capacity
    Material
    Membrane
    Pore size
    Sterile
    Availability
    Price
  • 77507-914
    Bottle top vacuum filter
    Bottle top vacuum filter
    50 ml
    Polystyrene
    PES
    0.22 µm
    +
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $191.59
    $191.59
    Case of 24
       $191.59
    Supplier Number: 347011
  • 77507-916
    Bottle top vacuum filter
    Bottle top vacuum filter
    50 ml
    Polystyrene
    PES
    0.10 µm
    +
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $191.59
    $191.59
    Case of 24
       $191.59
    Supplier Number: 347012
  • 77507-918
    Bottle top vacuum filter
    Bottle top vacuum filter
    50 ml
    Polystyrene
    PES
    0.45 µm
    +
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $191.59
    $191.59
    Case of 24
       $191.59
    Supplier Number: 347013
  • 77507-926
    Bottle top vacuum filter
    Bottle top vacuum filter
    50 ml
    Polystyrene
    PVDF
    0.22 µm
    +
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $202.17
    $202.17
    Case of 24
       $202.17
    Supplier Number: 347111
  • 77507-928
    Bottle top vacuum filter
    Bottle top vacuum filter
    50 ml
    Polystyrene
    PVDF
    0.10 µm
    +
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $202.17
    $202.17
    Case of 24
       $202.17
    Supplier Number: 347112
  • 77507-930
    Bottle top vacuum filter
    Bottle top vacuum filter
    50 ml
    Polystyrene
    PVDF
    0.45 µm
    +
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $202.17
    $202.17
    Case of 24
       $202.17
    Supplier Number: 347113
  • 77507-888
    Bottle top vacuum filter
    Bottle top vacuum filter
    150 ml
    Polystyrene
    PES
    0.22 µm
    +
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $179.61
    $179.61
    Case of 24
       $179.61
    Supplier Number: 346011
  • 77507-890
    Bottle top vacuum filter
    Bottle top vacuum filter
    150 ml
    Polystyrene
    PES
    0.10 µm
    +
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $179.61
    $179.61
    Case of 24
       $179.61
    Supplier Number: 346012
  • 77507-892
    Bottle top vacuum filter
    Bottle top vacuum filter
    150 ml
    Polystyrene
    PES
    0.45 µm
    +
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $179.61
    $179.61
    Case of 24
       $179.61
    Supplier Number: 346013
  • 77507-902
    Bottle top vacuum filter
    Bottle top vacuum filter
    150 ml
    Polystyrene
    PVDF
    0.22 µm
    +
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $182.48
    $182.48
    Case of 24
       $182.48
    Supplier Number: 346111
  • 77507-904
    Bottle top vacuum filter
    Bottle top vacuum filter
    150 ml
    Polystyrene
    PVDF
    0.10 µm
    +
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $182.48
    $182.48
    Case of 24
       $182.48
    Supplier Number: 346112
  • 77507-906
    Bottle top vacuum filter
    Bottle top vacuum filter
    150 ml
    Polystyrene
    PVDF
    0.45 µm
    +
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $182.48
    $182.48
    Case of 24
       $182.48
    Supplier Number: 346113
  • 77507-750
    Bottle top vacuum filter
    Bottle top vacuum filter
    250 ml
    Polystyrene
    PES
    0.22 µm
    +
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $184.01
    $184.01
    Case of 24
       $184.01
    Supplier Number: 342011
  • 77507-752
    Bottle top vacuum filter
    Bottle top vacuum filter
    250 ml
    Polystyrene
    PES
    0.10 µm
    +
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $184.01
    $184.01
    Case of 24
       $184.01
    Supplier Number: 342012
  • 77507-754
    Bottle top vacuum filter
    Bottle top vacuum filter
    250 ml
    Polystyrene
    PES
    0.45 µm
    +
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $184.01
    $184.01
    Case of 24
       $184.01
    Supplier Number: 342013
  • 77507-764
    Bottle top vacuum filter
    Bottle top vacuum filter
    250 ml
    Polystyrene
    PVDF
    0.22 µm
    +
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $191.59
    $191.59
    Case of 24
       $191.59
    Supplier Number: 342111
  • 77507-766
    Bottle top vacuum filter
    Bottle top vacuum filter
    250 ml
    Polystyrene
    PVDF
    0.10 µm
    +
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $191.59
    $191.59
    Case of 24
       $191.59
    Supplier Number: 342112
  • 77507-768
    Bottle top vacuum filter
    Bottle top vacuum filter
    250 ml
    Polystyrene
    PVDF
    0.45 µm
    +
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $191.59
    $191.59
    Case of 24
       $191.59
    Supplier Number: 342113
  • 77507-776
    Bottle top vacuum filter
    Bottle top vacuum filter
    500 ml
    Polystyrene
    PES
    0.22 µm
    +
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $222.11
    $222.11
    Case of 24
       $222.11
    Supplier Number: 343011
  • 77507-778
    Bottle top vacuum filter
    Bottle top vacuum filter
    500 ml
    Polystyrene
    PES
    0.10 µm
    +
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $222.11
    $222.11
    Case of 24
       $222.11
    Supplier Number: 343012
  • 77507-780
    Bottle top vacuum filter
    Bottle top vacuum filter
    500 ml
    Polystyrene
    PES
    0.45 µm
    +
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $222.11
    $222.11
    Case of 24
       $222.11
    Supplier Number: 343013
  • 77507-790
    Bottle top vacuum filter
    Bottle top vacuum filter
    500 ml
    Polystyrene
    PVDF
    0.22 µm
    +
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $237.16
    $237.16
    Case of 24
       $237.16
    Supplier Number: 343111
  • 77507-792
    Bottle top vacuum filter
    Bottle top vacuum filter
    500 ml
    Polystyrene
    PVDF
    0.10 µm
    +
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $237.16
    $237.16
    Case of 24
       $237.16
    Supplier Number: 343112
  • 77507-794
    Bottle top vacuum filter
    Bottle top vacuum filter
    500 ml
    Polystyrene
    PVDF
    0.45 µm
    +
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $237.16
    $237.16
    Case of 24
       $237.16
    Supplier Number: 343113
  • 77507-802
    Bottle top vacuum filter
    Bottle top vacuum filter
    1000 ml
    Polystyrene
    PES
    0.22 µm
    +
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $316.83
    $316.83
    Case of 24
       $316.83
    Supplier Number: 344011
  • 77507-804
    Bottle top vacuum filter
    Bottle top vacuum filter
    1000 ml
    Polystyrene
    PES
    0.10 µm
    +
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $316.83
    $316.83
    Case of 24
       $316.83
    Supplier Number: 344012
  • 77507-806
    Bottle top vacuum filter
    Bottle top vacuum filter
    1000 ml
    Polystyrene
    PES
    0.45 µm
    +
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $316.83
    $316.83
    Case of 24
       $316.83
    Supplier Number: 344013
  • 77507-816
    Bottle top vacuum filter
    Bottle top vacuum filter
    1000 ml
    Polystyrene
    PVDF
    0.22 µm
    +
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $324.30
    $324.30
    Case of 24
       $324.30
    Supplier Number: 344111
  • 77507-818
    Bottle top vacuum filter
    Bottle top vacuum filter
    1000 ml
    Polystyrene
    PVDF
    0.10 µm
    +
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $324.30
    $324.30
    Case of 24
       $324.30
    Supplier Number: 344112
  • 77507-820
    Bottle top vacuum filter
    Bottle top vacuum filter
    1000 ml
    Polystyrene
    PVDF
    0.45 µm
    +
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $324.30
    $324.30
    Case of 24
       $324.30
    Supplier Number: 344113