ContactUsLinkComponent
Variable Inertia Apparatus, Transparent
Variable Inertia Apparatus, Transparent
Catalog # 470356-478
Supplier:  GSC International
undefined
Variable Inertia Apparatus, Transparent
Catalog # 470356-478
Supplier:  GSC International
Supplier Number:  VBINR-CLR
Specifications
  • Description:
    Variable inertia apparatus, transparent
  • Cat. No.:
    470356-478
Specifications
About this item
Explore rotational acceleration with this pair of variable inertia disks.
  • Two disks allow students to compare the effects of different mass placements simultaneously
  • Semi-transparent plastic allows students to see masses inside
The Variable Inertia demonstrates an abstract concept in an easy-to-understand manner. It consists of two semi-clear plastic discs with the same mass and diameter. The discs are hollow inside with molded compartments. The 19 mm steel balls can be placed inside the discs in a variety of configurations. Based on the location of the steel balls, the distribution of the mass will change, and the discs will become either "center heavy" or "rim heavy".
Ask your students, how does changing the position of the mass affect the speed at which the discs roll? The semi-transparent discs will allow your student to see where the mass has been placed inside the discs. After conducting the experiment, the student will be able to relate the position of the mass to the outcome of the experiment and answer the question.