ContactUsLinkComponent
Anticon® SuperSorb Cleanroom Mop, Contec®
Anticon® SuperSorb Cleanroom Mop, Contec®
Catalog # 45800-080
Supplier:  Contec
Small Business Enterprise
undefined
Anticon® SuperSorb Cleanroom Mop, Contec®
Catalog # 45800-080
Supplier:  Contec
Supplier Number:  492238-649
Small Business Enterprise
Specifications
  • Description:
    Anticon® SuperSorb Cleanroom Mop
  • Cat. No.:
    45800-080
  • Supplier no.:
    492238-649
Specifications
About this item
Constructed from 100% continuous filament polyester with unique two-yarn system for superior sorbency
Designed for wet mopping in critical areas where maximum sorbency is required for applying cleaning and disinfectant solutions. Mop is durable and launderable for multiple uses. Designed for use with commercially available handles and bucket-wringer systems. Autoclavable for sterile applications.
Ordering information: Individually packed.