HelixMark® Standard Platinum-Cured Silicone Tubing, Nusil Material, Freudenberg Medical
Supplier: Freudenberg Medical
Certificates
About this item
HelixMark® platinum-cured silicone tubing meets the needs of a broad range of applications and is universally applicable for the transfer of fluids under standard requirements.
- Fully autoclavable and sterilizable
- Ideal for use in transfer operations as well as other applications requiring non-leachable material
- Material certification in every package
- Lot traceability
HelixMark® tubing meets USP Class VI and EP 3.1.9 biocompatiblity requirements. It is manufactured in Freudenberg Medical's FDA registered medical device facility following Good Manufacturing Practices (GMPs) in accordance with The Quality Systems Requlations.
Tubing is individually packaged in 50 foot coil. Durometer hardness: Shore A, 50.
Certifications: Certified to meet ISO 13485 quality standards.
Ordering information: Tubing is packaged double-bagged in crush-resistant boxes with material certification and lot traceability included in every box.