Perfluoro Acid Anhydrides
PI63164
:
- Description:Heptafluorobutyric acid anhydride reagent
- Size:10×1 ml
- Cat. No.:PI63164
- Supplier no.:TS-63164
Prepare fluoracyl derivatives for GC/MS and for flame ionization and electron-capture detection techniques with the Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA). These reagents react readily with alcohols, phenols, and amines.
- Used to prepare fluoracyl derivatives for GC-MS
- Storage at Room Temperature
- For use with GC/MS
- Standard components are Heptafluoro-butanoic acid anhydride
- Standard Concentration 98 to 100%
Produce stable volatile derivatives for FID and ECD techniques.