ContactUsLinkComponent
Homogenizer, T 25 easy clean digital
Homogenizer, T 25 easy clean digital
Catalog # 76329-872
Supplier:  IKA Works
undefined
Homogenizer, T 25 easy clean digital
Catalog # 76329-872
Supplier:  IKA Works
Supplier Number:  0025002561
Specifications
  • Description:
    T 25 easy clean digital homogenizer
  • Cat. No.:
    76329-872
Specifications
About this item
The T 25 easy clean digital is a high-performance dispersing instrument with electronic speed control from 3000 - 25000 rpm for working volumes of 1 - 2000 ml (H₂O).
  • No disassembly required
  • Brushless motor guarantees dust-free and silent operation
  • USB interface for control and documentation through labworldsoft®
  • Timer/counter function
  • Increased safety through tool recognition and IP-class
The S 25 dispersing elements allows the T 25 to be used in various applications- whether it is homogenizing medical samples, emulsifying cosmetics or the suspension of coatings.
In combination with reactor it is also possible to conduct dispersion tasks under vacuum or pressure.