ContactUsLinkComponent
ProPac™ 3R SAX 3 μm HPLC Columns
ProPac™ 3R SAX 3 μm HPLC Columns
  • Catalog No:
  • 76748-626
  • 76748-624
  • 76748-620
  • 76748-622
  • Max. Pressure:
  • 4500 psi
  • 4500 psi
  • 4500 psi
  • 4500 psi
  • pH Range:
  • 2 to 12
  • 2 to 12
  • 2 to 12
  • 2 to 12
  • Temperature Limit:
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
Frequently Bought Together