ProPac™ 3R SCX 3 μm HPLC Columns
Supplier: Thermo Scientific
Certificates
About this item
Use Thermo Scientific™ ProPac™ 3R SCX 3 μm HPLC columns to provide the reproducibility, resolution, and robustness needed for protein characterization. These columns are ideal for charge variant analysis and recommended for use with therapeutic proteins, monoclonal antibodies (mAbs), antibody drug conjugates (ADCs), and biosimilar proteins.
Designed with bio-inert materials to reduce secondary interactions, our ProPac 3R SCX columns deliver the performance you need over a broad range of pH's, temperatures, and mobile phase compositions with excellent recovery and low carryover.
Specifications
- Catalog No:
- 76748-612
- 76748-614
- 76748-616
- 76748-618
- Max. Pressure:
- 4500 psi
- 4500 psi
- 4500 psi
- 4500 psi
- pH Range:
- 2 to 12
- 2 to 12
- 2 to 12
- 2 to 12
- Temperature Limit:
- 60 °C
- 60 °C
- 60 °C
- 60 °C
Specifications
Frequently Bought Together