About this item
Security units provide safe, reliable storage
Intermediate shelves can be mixed and matched from MetroMax i (all polymer) or MetroMax Q (polymer mats on epoxy coated steel frames). MetroMax i shelves are available with solid or open grid polymer mats. Metromax Q shelves are available with open grid mats and feature a quick release corner lever for ease of adjustability. Both styles feature removable polyropylene shelf mats for easy cleaning; mats are dishwasher safe. Microban™ antimicrobial product protection is built into all shelf mats to minimize growth of bacteria and fungus. Shelf mats are blue in color.
Available in three lengths.
Standard units inlcude top and bottom wire shelves, tri-lobal adapters, steel posts wire enclosures and doors. Mounted on four 12.7cm (5") diameter swivel casters with 14cm (51/2") bumpers. Intermediate shelves are sold separately.