RPR Syphilis Test Kits, ASI
Supplier: Arlington Scientific
Certificates
About this item
The ASI RPR Card Test is a screening test for the sexually transmitted disease, SYPHILIS. The disease is found worldwide and may be fatal if left untreated. To keep the blood supply safe, transfusion blood is tested for syphilis, HIV and hepatitis.
- A CDC recommended primary screening test
- Simple and economical to use
- Easy to interpret
- All testing materials provided
- Sold with controls or without
- 3 levels of liquid controls included: reactive, weak reactive, nonreactive
- Color-coded dropper vials
- Warp resistant coated test cards
- Screw cap vials - No glass ampules
- CPD, CPDA-1, EDTA, Heparin or Sodium Citrate may be used
- Made with pride in the USA
- Longest shelf life - up to 24 months
- Proven results on CAP, AAB, and API Surveys
- 10 and 30 well test cards available
- Sensitivity : >98%
- Specificity: >99%
Certifications: CE marked. CPT Code 86592. 510(k)-K851504
Frequently Bought Together