C-Reactive Protein (CRP) Slide Test Kits, ASI
Supplier: Arlington Scientific
Certificates
About this item
The ASI CRP Slide Test is a convenient and specific slide agglutination assay for the qualitative and semiquantitative detection of C-Reactive Protein (CRP) in human serum. No initial dilution of patient sample is required for this test.
- 2 Minute slide test
- No initial dilution required
- 2 Liquid controls included
- Longest dating - up to 24 month shelf life
- Proudly made in USA
- Proven by AAB, CAP and API surveys
- Sensitivity: 6 mg/L
- Specificity: >95%
Certifications: CE marked. CPT Code 86140. 510(k)-K821388
Frequently Bought Together