Specifications
- Size:#5
- Cat. No.:25856-023
- Supplier no.:371050
Specifications
About this item
This durable handle accommodates No
10, 11, 12, or 15 Bard-Parker® Rib-Back™ blades. Handle is shaped to provide a firm grip for entire palm of hand.