About this item
For use with No
3, No. 5, or No. 7 Bard-Parker® scalpel handles. Fine blade carbon steel. The Rib-Back™ prevents weaving and ensures greater accuracy in the line of cut. Specially ground and carefully inspected. Individually wrapped.