Adenosine-5'-triphosphate disodium salt (ATP disodium salt) trihydrate
Supplier: RPI
Certificates
About this item
CAS Number:51963-61-2
:987-65-5
Formula:C₁₀H₁₄N₅Na₂O₁₃P₃·3H₂O
MDL Number:MFCD00150754
:605.19 g/mol
Storage Temperature:Freezer
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Appearance:Fine white crystalline powder
- A250/A260:0.75 - 0.81
- A280/A260:0.14 - 0.18
- Heavy Metals:≤ 10 ppm
- Identification:Pass Test
- Moisture:≤ 10.0 %
- Molar Extinction:≥ 14700
- Purity:≥ 98.0 %
- Sodium (Flame Photom):7.0 - 8.0 %
- Solubility:Clear colourless and haze free
- Residual Solvents:Conforms
- Melamine Free:Melamine Free
- BSE/TSE Free:BSE/TSE Free
- Country of Origin:Report
Specification Test Results
Frequently Bought Together