ContactUsLinkComponent
Accessories for Synthware Solvent Distilling Head, Kemtech America
  60005-140
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Accessories for Synthware Solvent Distilling Head, Kemtech America
  60005-140
 :  S211944
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
  • Description:
    Replacement 2 mm Double Oblique PTFE Plug
  • Cat. No.:
    60005-140
  • Supplier no.:
    S211944
Kemtech replacement t-bore stopcock.
  • Borosilicate 3.3
  • Double Oblique
  • 1:5 Taper
Double Oblique, Teflon®, stopcock is a direct replacement for a number of Kemtech products.