CD-Chex CD34™ Flow Cytometry Controls, Streck
About this item
CD-Chex CD34™ is a positive procedural control for enumerating CD34 positive cells by flow cytometry
The CD34 positive cells have characteristics of human hematopoietic progenitor cells. CD-Chex CD34 is available in three levels, providing reference values similar to levels found in normal peripheral blood, bone marrow, cord blood and mobilized patient samples. It is compatible with ISHAGE gating strategies as well as commercially available enumeration kits. CD-Chex CD34 has 90-day closed-vial stability and 30-day open-vial stability.
5 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
CD-Chex CD34™ is a positive procedural control for enumerating CD34 positive cells by flow cytometry
The CD34 positive cells have characteristics of human hematopoietic progenitor cells. CD-Chex CD34 is available in three levels, providing reference values similar to levels found in normal peripheral blood, bone marrow, cord blood and mobilized patient samples. It is compatible with ISHAGE gating strategies as well as commercially available enumeration kits. CD-Chex CD34 has 90-day closed-vial stability and 30-day open-vial stability.
Product Family Options
Product Information
- SizeAvailabilityPrice
- 87005-266Level 1Supplier Number: 213337Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- 87005-268Levels 1 and 2Supplier Number: 213338Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- 87005-270Levels 1 and 2Supplier Number: 213339Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- 87005-276Level 2Supplier Number: 213347Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- 82030-694Level 3Supplier Number: 213383Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.