Orders LinkContactUsLinkComponent
Para 12® Plus Controls, Streck
Supplier:   Streck Laboratories
Para 12® Plus Controls, Streck
Certificates
Para 12® Plus Controls, Streck
Para 12® Plus Controls, Streck
6 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
A hematology control with assay values provided for the five-part white cell differential
Open-vial stability is seven days and closed-vial stability is 75 days. Packaged in a vial with pierceable cap. Compatible with Abbott® instruments.
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 87004-512
    Low Level
    Low Level
    3.0 mL
    Log in for availability
    Log in for availability
    Pack of 4 (3ml)
    Pack of 4 (3ml) - $210.05
    $210.05
    Pack of 4 (3ml)
       $210.05
    Supplier Number: 218924
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 87004-510
    Normal Level
    Normal Level
    3.0 mL
    Log in for availability
    Log in for availability
    Pack of 4 (3ml)
    Pack of 4 (3ml) - $209.02
    $209.02
    Pack of 4 (3ml)
       $209.02
    Supplier Number: 218914
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 87004-514
    High Level
    High Level
    3.0 mL
    Log in for availability
    Log in for availability
    Pack of 4 (3ml)
    Pack of 4 (3ml) - $210.05
    $210.05
    Pack of 4 (3ml)
       $210.05
    Supplier Number: 218934
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 87004-504
    1 Low, 1 Normal, and 1 High Levels
    1 Low, 1 Normal, and 1 High Levels
    3.0 mL
    Log in for availability
    Log in for availability
    Pack of 3 (3ml)
    Pack of 3 (3ml) - $191.74
    $191.74
    Pack of 3 (3ml)
       $191.74
    Supplier Number: 218903
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 87004-506
    2 Low, 2 Normal, and 2 High Levels, with Disk
    2 Low, 2 Normal, and 2 High Levels, with Disk
    3.0 mL
    Log in for availability
    Log in for availability
    Pack of 6 (3ml)
    Pack of 6 (3ml) - $350.07
    $350.07
    Pack of 6 (3ml)
       $350.07
    Supplier Number: 218907
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 87004-508
    4 Low, 4 Normal, and 4 High Levels, with Disk
    4 Low, 4 Normal, and 4 High Levels, with Disk
    3.0 mL
    Log in for availability
    Log in for availability
    Pack of 12 (3ml)
    Pack of 12 (3ml) - $592.52
    $592.52
    Pack of 12 (3ml)
       $592.52
    Supplier Number: 218913
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.