Orders LinkContactUsLinkComponent
Retic-Chex® Linearity Controls, Streck
Supplier:   Streck Laboratories
Retic-Chex® Linearity Controls, Streck
Certificates
Retic-Chex® Linearity Controls, Streck
Retic-Chex® Linearity Controls, Streck
1 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Controls are designed to establish the patient reportable range and the analytic linearity of the system
Retic-Chex stain is buffered and filtered new methylene blue that can be used with control and patient samples. It is packaged in a 3 mL plastic squeeze dropper vial and can be stored at room temperature.
Available in five-vial sets with values assigned. Open-vial stability is five days and closed-vial stability is 75 days. Compatible with Abbott®, Beckman Coulter®, and Sysmex® instruments.
 : Each control is supplied with five vials, one each for levels 1, 2, 3, 4 and 5. The 3.0 mL control is designed specifically for Beckman Coulter® LH 750/LH 755 instruments.
Product Family Options
Product Information
  • Size
    Pack type
    Availability
    Price
  • 82030-690
    Retic-Chex® Linearity Controls
    Retic-Chex® Linearity Controls
    2.5 mL
    Plastic Pierceable Vials
    Discontinued
    Supplier Number: -
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 82028-952
    Retic-Chex® Linearity Controls
    Retic-Chex® Linearity Controls
    3.0 mL
    Plastic Pierceable Vials
    Log in for availability
    Log in for availability
    Each
    Each - $674.80
    $674.80
    Each
       $674.80
    Supplier Number: 285535
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.