Chemical-Resistant Cleanroom Labels, Dot, CleanMark®, LG INTERNATIONAL
Supplier: Lgi
Certificates
About this item
Cleanroom Labels recommended for use in any cleanroom environment.
- Manufactured from 100% synthetic
- Non particulating
- Ultra-low outgassing raw materials
These removable labels can be easily removed without leaving behind adhesive residue contamination. Labels can be printed upon with a cleanroom pen or by any standard thermal transfer printer.
CleanMark® labels can be custom constructed to fit any size. These labels can withstand extreme hot or cold temperatures, harsh chemical solvents, and physical abrasion. They can indicate sterilization processes such as autoclaving, may be pre-sterilized, can firmly adhere to or easily peel from challenging surfaces, and will not contaminate clean or sterile environments with particulates, outgassing, or microbes. Please call your Avantor Sales Representative for additional information.
Certifications: RoHs compliant.
Frequently Bought Together