Orders LinkContactUsLinkComponent
[beta]-Amyloid Peptide (1-42)
250 µg
  80500-030
 :  PP69-.25MG
[beta]-Amyloid Peptide (1-42)
  80500-030
 :  
[beta]-Amyloid Peptide (1-42)
250 µg
 :  
$523.14
Each (250µG)
Synthetic peptide corresponding to amino acids 1–42 of the processed human amyloid peptide. Supplied in a form that is not neurotoxic prior to preincubation. The level of toxicity has recently been shown to correlate to the extent of beta sheet structure. Reconstitute with degassed HPLC grade deionized water.
  • Size:
    250 µg
  • Cat. No.:
    80500-030
  • Protein/Peptide Name:
    [beta]-Amyloid Peptide (1-42)