Order Entry
United States
ContactUsLinkComponent
BCECF AM ester ≥85% (by HPLC), Calbiochem®, Sigma-Aldrich®
BCECF AM ester ≥85% (by HPLC), Calbiochem®, Sigma-Aldrich®
Catalog # 80050-710
Supplier:  MilliporeSigma
CAS Number:  
BCECF AM ester ≥85% (by HPLC), Calbiochem®, Sigma-Aldrich®
Catalog # 80050-710
Supplier:  MilliporeSigma
Supplier Number:  216254-1MG
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 mg
  • Cat. No.:
    80050-710
  • Supplier no.:
    216254-1MG

Specifications

About this item

 :
 :808.69 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Freezer
CAS Number:117464-70-7
Formula:C₄₂H₄₀O₂₁
MDL Number:MFCD00036969