Trolox, EMD Millipore
Catalog # 80057-672
Supplier: MilliporeSigma
CAS Number:
Specifications
- Size:500 mg
- Cat. No.:80057-672
- Supplier no.:648471-500MG
Specifications
About this item
A cell-permeable, water-soluble derivative of vitamin E with potent antioxidant properties
Prevents peroxynitrite-mediated oxidative stress and apoptosis in rat thymocytes.