Biohazard Warning Label, National Marker
Catalog # 36434-434
Supplier: ACCUFORM MANUFACTURING, INC
Specifications
- Description:Biohazard warning label
- Dimensions:76.2×76.2 mm (3×3")
- Material:Plastic
- Cat. No.:36434-434
- Supplier no.:S52RL
Specifications
About this item
Displays biohazard symbol and word.
- Pressure-sensitive paper rolls can be written on to provide additional information
Ordering information: Each roll contains 500 individual labels.