

Guanidinium thiocyanate
Supplier: RPI
Certificates
About this item
CAS Number:593-84-0
:
Formula:CH₅N₃·CHNS
MDL Number:MFCD00013027
:118.16 g/mol
EINECS:209-812-1
:GHS05
:GHS07
Storage Temperature:Room Temperature
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Appearance:White to slightly off white crystalline powder
- Assay (Argentometric Titration):≥ 99.0 %
- Assay (Ion Chromatography):≥ 99.5 %
- Chloride Content:≤ 1.0 %
- Cobalt (Co):≤ 5 ppm
- FTIR:Conforms to reference
- Iron:≤ 5 ppm
- pH:5.0 - 7.0
- DNase/ RNase:None Detected
- Melting Point:118 - 121 oC
- Solubility:Pass
- Sulfate Ash:≤ 0.5 %
- Moisture:≤ 0.5 %
- BSE/TSE Free:BSE/TSE Free
- Grade:Ultra Pure
- Country of Origin:Report
Specification Test Results
Frequently Bought Together