Vacuum Cup Kit, Virtual Industries
21925-166
- Description:Vacuum Cup Kit
- Cat. No.:21925-166
- Supplier no.:V9000K8-B
High-quality ESD-safe vacuum cups for use in electronics/SMT, optics, mechanical assembly, semiconductor handling and robotics
Made of black Buna-n static dissipative material.
: Supplied with one each of the following seven diameters: 3/32", 1/8", 1/4", 3/8", 1/2", 5/8", 3/4".