HANDI-VAC® Vacuum Handling System, Virtual Industries
Supplier: Virtual Industries
Certificates
About this item
This low-cost vaccum handling system works like a tweezer
Tool features a squeeze bulb design and is made of black static-dissipative rubber. Required vacuum cups are available in two sizes for various handling needs. Vaccum cups are constructed of clear high-temperature silicone and can attach directly to the tool or be expanded via the metal probe.
Frequently Bought Together