Halt™ Protease and Phosphatase Inhibitor Cocktails, Thermo Scientific
Halt Protease and Phosphatase Inhibitor Cocktail (100X) provides the convenience of a single solution with full protein sample protection for cell and tissue lysates.
- 100X liquid formulation is easier and more effective to use than individual inhibitors
- Aqueous-based format allows for convenient cold-room storage
- Free of AEBSF and can be used in Mass spectrometry
- Compatible with Thermo Scientific Pierce Cell Lysis Buffers
This Halt Protease and Phosphatase Inhibitor Cocktail is specifically optimized to protect proteins from degradation during extraction and purification. The cocktail contains inhibitors against the major classes of proteases and phosphatases, targeting aminopeptidases, cysteine and serine proteases and serine/threonine and protein tyrosine phosphatases.
- Storage Conditions:Upon receipt store at 4˚C. Product shipped with an ice pack.
Frequently Bought Together