Order Entry
United States
ContactUsLinkComponent
 
 
 
Estradiol ≥97%, Calbiochem®, Sigma-Aldrich®
  80056-424
 :  
 
 
Estradiol ≥97%, Calbiochem®, Sigma-Aldrich®
  80056-424
 :  3301-1GM
 :  

 

  • Pack type:
    Glass bottle
  • Size:
    1 g
  • Cat. No.:
    80056-424
  • Supplier no.:
    3301-1GM

 

 

 :
 :272.39 g/mol
 :M-Clarity™ quality level = MQ100
 :200-023-8
 :GHS08
 :GHS09
 :Refrigerator
 :50-28-2
 :C₁₈H₂₄O₂
 :MFCD00003693