beta-Galactosidase (from E. coli)
Catalog # RL-0000-17
Supplier: Rockland Immunochemical
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Specifications
- Source:E. coli
- Size:1 mg
- Enzyme Name:beta-Galactosidase
- Cat. No.:RL-0000-17
- Supplier no.:0000-17
Specifications
About this item
All enzymes and related reagents are configured as an integrated system and have been thoroughly tested.
Application: Control in immunological assays.