Order Entry
United States
ContactUsLinkComponent
Trypsin, Gold
Trypsin, Gold
Catalog # PAV5280
CAS Number:  
undefined
Trypsin, Gold
Catalog # PAV5280
Supplier Number:  V5280
CAS Number:  

Specifications

  • Conjugation:
    Gold
  • Size:
    100 μg
  • Storage Conditions:
    –30...–10 °C
  • Enzyme Name:
    Trypsin
  • Grade:
    Mass Spectrometry grade
  • Cat. No.:
    PAV5280
  • Supplier no.:
    V5280

Specifications

About this item

Trypsin Gold, Mass Spectrometry Grade, provides maximum specificity for mass spectrometry.

  • Maximum Digest Specificity with Extreme Resistance to Autolytic Digestion
  • Each lot qualified by mass spectrometry to ensures compatibility with your applications/instrumentation
  • Chymotrypsin activity eliminated for distinct, consistent data
  • Digest in gel, or in solution

Trypsin is a serine protease that specifically cleaves at the carboxylic side of lysine and arginine residues. The stringent specificity of trypsin is essential for protein identification. Native trypsin is subject to autolysis, generating pseudotrypsin, which exhibits a broadened specificity including a chymotrypsin-like activity. Such autolysis products, present in a trypsin preparation, would result in additional peptide fragments that could interfere with database analysis of the mass of fragments detected by mass spectrometry. Trypsin Gold, Mass Spectrometry Grade, has been manufactured to provide maximum specificity. Lysine residues in the porcine trypsin have been modified by reductive methylation, yielding a highly active and stable molecule that is extremely resistant to autolytic digestion. The specificity of the purified trypsin is further improved by TPCK treatment, which inactivates chymotrypsin. The treated trypsin is then purified by affinity chromatography and lyophilized to yield Trypsin Gold, Mass Spectrometry Grade. It is resistant to mild denaturing conditions such as 0.1% SDS, 1M urea or 10% acetonitrile and retains 50% of its activity in 2M guanidine HCl. The activity of trypsin is decreased when acidic residues are present on either side of a susceptible bond. If proline is at the carboxylic side of lysine or arginine, the bond is almost completely resistant to cleavage. Each lot of quality-tested Trypsin Gold, Mass Spectrometry Grade, is qualified for use with in-gel digestion and mass spectrometric analysis.

Each lot is quality tested and qualified for use with in-gel digestion and mass spectrometric analysis. The trypsin is provided in one vial, and stability is ensured up to five freeze-thaw cycles.

Native trypsin is subject to autolysis, which broadens its specificity and results in additional peptide fragments that could interfere with database analysis on fragments determined in mass spectrometry. It has been manufactured to provide maximum specificity. Lysine residues in the porcine trypsin have been modified by reductive methylation, yielding a highly active and stable molecule that is resistant to autolytic digestion. The specificity is further improved by TPCK treatment to inactivate any chymotrypsin present. The treated trypsin is then purified by affinity chromatography and lyophilized.