Seals, Optimize Technologies
About this item
Piston seals are designed to offer reliable performance in a variety of operating conditions.
- Polyethylene-based seals are designed for applications where mostly aqueous or buffered-aqueous mobile phases are used
- Graphetized PTFE seals are designed for applications where mobile phases are mostly organic or where chemical compatibility is of primary concern
- Reliable performance in a variety of operating conditions
Installing a piston seal can be a tricky undertaking. A relatively small seal must be fit into the seal cavity in such a way to ensure that it is pressed in evenly without distortion or damage. The Optimize Seal Forming/ Insertion Tool Kit was specifically designed to aid in this endeavor. This kit contains three tools to accommodate all of your HPLC/ UHPLC piston seals. One end has a beveled cavity for seal forming, while the other is an insertion support for perfect seal installation. Also included with each kit are complete instructions for maximizing seal life and a solvent compatibility guide to aid in prudent seal material selection.
OPTI-SEAL® UHMW-PE polyethylene-based seals are designed for applications where mostly aqueous or buffered-aqueous mobile phases are used. ITB™ graphetized PTFE seals are designed for applications where mobile phases are mostly organic or where chemical compatibility is of primary concern.
290 Options Available Below
Product Details & Documents
Piston seals are designed to offer reliable performance in a variety of operating conditions.
- Polyethylene-based seals are designed for applications where mostly aqueous or buffered-aqueous mobile phases are used
- Graphetized PTFE seals are designed for applications where mobile phases are mostly organic or where chemical compatibility is of primary concern
- Reliable performance in a variety of operating conditions
Installing a piston seal can be a tricky undertaking. A relatively small seal must be fit into the seal cavity in such a way to ensure that it is pressed in evenly without distortion or damage. The Optimize Seal Forming/ Insertion Tool Kit was specifically designed to aid in this endeavor. This kit contains three tools to accommodate all of your HPLC/ UHPLC piston seals. One end has a beveled cavity for seal forming, while the other is an insertion support for perfect seal installation. Also included with each kit are complete instructions for maximizing seal life and a solvent compatibility guide to aid in prudent seal material selection.
OPTI-SEAL® UHMW-PE polyethylene-based seals are designed for applications where mostly aqueous or buffered-aqueous mobile phases are used. ITB™ graphetized PTFE seals are designed for applications where mobile phases are mostly organic or where chemical compatibility is of primary concern.
Product Family Options
DION
Product Information
- For use withAvailabilityPrice
- 77100-618OPTI-SEAL UHMW-PE Piston Seal, (Elastomer O-Ring)OPTI-SEAL UHMW-PE Piston Seal, (Elastomer O-Ring)Dionex Analytical PumpsLog in for availabilityEach$130.37Supplier Number: 25-36-00599
- 21510-448OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, PEEK CartridgeDionex DQP, GPM-1, GPM-2, DXP, AGP (Analytical), DRP, 2000, 4000, SP-1, DP-1, ICS-3000, ICS-5000 (Analytical Version Only)Log in for availabilityEach$664.17Supplier Number: 25-59-00613
- 21510-460OPTI-SEAL UHMW-PE Piston Seal (Hastelloy C Spring)OPTI-SEAL UHMW-PE Piston Seal (Hastelloy C Spring)Dionex/All AnalyticalLog in for availabilityEach$166.52Supplier Number: 25-36-00597
- 21510-462OPTI-SEAL UHMW-PE Piston Seal (Hastelloy C Spring)OPTI-SEAL UHMW-PE Piston Seal (Hastelloy C Spring)Dionex Analytical PumpsLog in for availabilityPack of 10$1,665.23Supplier Number: 25-36-00598
- 21510-538OPTI-MAX Outlet Check Valve (New Style), 1/8" Ruby Ball/Sapphire Seat, PEEK CartridgeOPTI-MAX Outlet Check Valve (New Style), 1/8" Ruby Ball/Sapphire Seat, PEEK CartridgeDionex GPM-2,DXP,AGP (Analytical), 2000, 4000, SP-1, DP-1, ICS-3000, ICS-5000 (Analytical Version Only)Log in for availabilityEach$664.17Supplier Number: 25-59-00612
- 77100-620OPTI-SEAL UHMW-PE Piston Seal, (Elastomer O-Ring)OPTI-SEAL UHMW-PE Piston Seal, (Elastomer O-Ring)Dionex Analytical PumpsLog in for availabilityPack of 10$1,304.48Supplier Number: 25-36-00600
- 77100-622Back-Up SealSupplier Number: 25-36-00601
- 77100-624Back-Up SealSupplier Number: 25-36-00602
- 77100-628OPTI-MAX Outlet Check Valve (New Style), 1/8" Ceramic Ball & Seat, PEEK CartridgeOPTI-MAX Outlet Check Valve (New Style), 1/8" Ceramic Ball & Seat, PEEK CartridgeDionex GPM-2, DXP, AGP 2000, 4000, SP-1, DP-1, ICS-3000, ICS-5000 (Analytical Versions Only)Log in for availabilityEach$657.72Supplier Number: 25-58-00612
- 77100-630OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK CartridgeDionex DQP, GPM-1, GPM-2, DXP, AGP, DRP, 2000, 4000, SP-1, DP-1, ICS-3000, ICS-5000 (Analytical Version Only)Log in for availabilityEach$657.72Supplier Number: 25-58-00613
- 83008-324Deuterium LampSupplier Number: 25-32-02081
- 83008-326Deuterium LampSupplier Number: 25-32-02820
- 83008-476OPTI-MAX Cartridge Check Valve Conversion KitOPTI-MAX Cartridge Check Valve Conversion KitDionex GPM-2, XP, 2000, 4000, SP-1, DP-1, ICS-3000, ICS-5000 (Analytical Version Only)Log in for availabilityEach$1,336.36Supplier Number: 25-58-02143
- Supplier Number: 25-34-04543
HP/Agilent
Product Information
- For use withAvailabilityPrice
- 77100-664Sapphire PlungerSupplier Number: 27-34-00658
- 77100-672OPTI-SEAL UHMW-PE Plunger SealSupplier Number: 27-36-01570
- 77100-674OPTI-SEAL UHMW-PE Plunger SealOPTI-SEAL UHMW-PE Plunger SealHP/Agilent 1050, 1100, 1200Log in for availabilityPack of 10$1,843.36Supplier Number: 27-36-01571
- 77100-676ITB PTFE Plunger SealSupplier Number: 27-36-01572
- 77100-678ITB PTFE Plunger SealSupplier Number: 27-36-01573
- 77100-680OPTI-SEAL UHMW-PE Wash SealSupplier Number: 27-36-02364
- 77100-684OPTI-SEAL UHMW-PE Wash SealSupplier Number: 27-36-02365
- 77100-686ITB PTFE Wash SealSupplier Number: 27-36-02366
- 77100-690ITB PTFE Wash SealSupplier Number: 27-36-02367
- 83008-342Seal Cap, 2-in-1Seal Cap, 2-in-1Agilent 1100, 1050, 1200 (replaces OEM Gold Seal and Outlet Cap)Log in for availabilityEach$82.72Supplier Number: 27-31-02853
- 83008-348Deuterium LampDeuterium LampAgilent/HP C Series, 1050 C, 1050 DAD, 1090, 1040, 79583CLog in for availabilityEach$1,476.62Supplier Number: 27-32-02844
- 83008-350Deuterium DoubleLife LampSupplier Number: 27-32-02845
- 83008-352Deuterium DoubleLife LampDeuterium DoubleLife LampAgilent/HP 1100, 1200 VWDDiscontinued-Supplier Number: -
- 83008-354Gasket Wash SealSupplier Number: 27-36-02851
- 89522-328PTFE FritsPTFE FritsAgilent 1050, 1100, 1200Discontinued-Supplier Number: 27-04-02852
WATE
Product Information
- For use withAvailabilityPrice
- Discontinued-Supplier Number: -
- Discontinued-Supplier Number: -
- Discontinued-Supplier Number: -
- Discontinued-Supplier Number: -
- 21509-899OPTI-MAX EF Inlet Cartridge Check Valve 3/16" Ruby Ball & Sapphire Seat, PEEK CartridgeOPTI-MAX EF Inlet Cartridge Check Valve 3/16" Ruby Ball & Sapphire Seat, PEEK CartridgeWaters 225 µL (#3) [510, 590, 600/600E, 610, 650, 6K/6KA]Log in for availabilityEach$788.01Supplier Number: 39-69-01262
- 21509-901OPTI-MAX EF Outlet Cartridge Check Valve 3/16" Ruby Ball & Sapphire Seat, PEEK CartridgeOPTI-MAX EF Outlet Cartridge Check Valve 3/16" Ruby Ball & Sapphire Seat, PEEK CartridgeWaters 400 µL (#4) & 900 µL (#6) Heads [Delta Prep 3000, 600/600E, 650]Log in for availabilityEach$788.01Supplier Number: 39-69-01267
- 21510-269Needle w/ Nut & FerruleSupplier Number: 39-31-01147
- 21510-294U6K Injector SealSupplier Number: 39-30-01124
- 21509-068Ruby Ball & SeatSupplier Number: 39-38-01308
- 21509-1023/16" Ruby Ball & SeatSupplier Number: 39-38-01311
- 21509-134ITB PTFE Plunger SealITB PTFE Plunger SealWaters 225 µL Head (#3) [510, 590, 600/600E, 610, 650, 6K/6KA]Log in for availabilityEach$119.05Supplier Number: 39-36-01214
- 21509-136ITB PTFE Plunger SealITB PTFE Plunger SealWaters 225 µL Head (#3)[510, 590, 600/600E, 610, 650, 6K/6KA]Log in for availabilityPack of 10$1,241.31Supplier Number: 39-36-01215
- 46600-640ITB PTFE Plunger Seal, BlackITB PTFE Plunger Seal, BlackWaters 100 µL Head [M45/45G, M501, 510, 590, 600/600E, 610, 6K/6KA, 515, 1515, 1525], 10/PkLog in for availabilityPack of 10$1,341.51Supplier Number: 39-36-01207
- 46600-642PlungerSupplier Number: 39-34-01188
- 77101-164Waters Type Union w/ Nuts & Ferrules for 1/16" OD TubingSupplier Number: 39-24-01088
- 77101-166Waters Type Compression Screw for 1/16" OD TubingSupplier Number: 39-24-01090
- 77101-168Waters Type Compression Screws & Ferrules, (39-24-01090 & 39-24-01291), 10/PkWaters Type Compression Screws & Ferrules, (39-24-01090 & 39-24-01291), 10/PkWATELog in for availabilityPack of 10$392.16Supplier Number: 39-24-01091
- 77101-170Waters Type PEEK Ferrule for 1/16" OD TubingSupplier Number: 39-24-01290
- 77101-172Waters Type SS Ferrule for 1/16" OD TubingSupplier Number: 39-24-01291
- 77101-192WISP Solvent SealSupplier Number: 39-30-01113
- 77101-210Priming Syringe Kit, Contains Priming Syringe & Needle, Nut & FerrulePriming Syringe Kit, Contains Priming Syringe & Needle, Nut & FerruleWATELog in for availabilityEach$678.16Supplier Number: 39-31-01151
- 77101-218Optimize Safety SyringeSupplier Number: 39-31-01171
- 77101-238PlungerSupplier Number: 39-34-01191
- 77101-246PlungerPlungerWaters Alliance/2690, 2695, 2790, 2795Discontinued-Supplier Number: 39-34-02180
- 77101-250ITB PTFE Plunger Seal, BlackITB PTFE Plunger Seal, BlackWaters 100 µL Head [M45/45G, M501, 510, 590, 600/600E, 610, 6K/6KA, 515, 1515, 1525]Log in for availabilityEach$148.77Supplier Number: 39-36-01206
- 77101-254OPTI-SEAL UHMW-PE Plunger SealOPTI-SEAL UHMW-PE Plunger SealWaters 225 µL Head (#3)[510, 590, 600/600E, 610, 650, 6K/6KA]Log in for availabilityEach$109.55Supplier Number: 39-36-01216
- 77101-256OPTI-SEAL UHMW-PE Plunger SealOPTI-SEAL UHMW-PE Plunger SealWaters 225 µL Head (#3) [510, 590, 600/600E, 610, 650, 6K/6KA]Log in for availabilityPack of 10$1,141.09Supplier Number: 39-36-01217
- 77101-258OPTI-SEAL UHMW-PE Plunger SealOPTI-SEAL UHMW-PE Plunger SealWaters 100 µL Head [M45/45G, M501, 510, 590, 600/600E, 610, 6K/6KA, 515, 1515, 1525]Log in for availabilityEach$136.89Supplier Number: 39-36-01305
- 77101-260OPTI-SEAL UHMW-PE Plunger SealOPTI-SEAL UHMW-PE Plunger SealWaters 100 µL Head [M45/45G, M501, 510, 590, 600/600E, 610, 6K/6KA, 515, 1515, 1525]Log in for availabilityPack of 10$1,414.95Supplier Number: 39-36-01306
- 77101-262OPTI-SEAL UHMW-PE Plunger SealSupplier Number: 39-36-02124
- 77101-264ITB PTFE Plunger SealSupplier Number: 39-36-02127
- 77101-266OPTI-SEAL UHMW-PE Plunger SealSupplier Number: 39-36-02228
- 77101-268ITB PTFE Plunger SealSupplier Number: 39-36-02229
- 77101-270OPTI-SEAL UHMW-PE Plunger SealOPTI-SEAL UHMW-PE Plunger SealWaters Alliance/2690, 2695, 2790, 2795Log in for availabilityEach$124.55Supplier Number: 39-36-02596
- 77101-272OPTI-SEAL UHMW-PE Plunger SealOPTI-SEAL UHMW-PE Plunger SealWaters Alliance/2690, 2695, 2790, 2795Log in for availabilityPack of 10$1,249.32Supplier Number: 39-36-02597
- 77101-294OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, SS CartridgeWaters 616, AllianceLog in for availabilityEach$652.20Supplier Number: 39-46-01949
- 77101-302OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, SS CartridgeWaters 616, AllianceLog in for availabilityEach$656.07Supplier Number: 39-47-01949
- 77101-304OPTI-MAX Outlet Cartridge Check Valve, 1/16" Ruby/Sapphire Seat, SS CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/16" Ruby/Sapphire Seat, SS CartridgeWaters 616, AllianceLog in for availabilityEach$656.07Supplier Number: 39-47-01950
- 77101-310OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ceramic Ball/Ceramic Seat, PEEK Cartridge, BiocompatibleOPTI-MAX Inlet Cartridge Check Valve, 1/16" Ceramic Ball/Ceramic Seat, PEEK Cartridge, BiocompatibleWaters 625/626Log in for availabilityEach$610.70Supplier Number: 39-48-01907
- 77101-312OPTI-MAX Outlet Cartridge Check Valve, 1/16" Ceramic Ball/Ceramic Seat, PEEK Cartridge, BiocompatibleOPTI-MAX Outlet Cartridge Check Valve, 1/16" Ceramic Ball/Ceramic Seat, PEEK Cartridge, BiocompatibleWaters 625/626Log in for availabilityEach$622.25Supplier Number: 39-48-01909
- 77101-314OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ceramic Ball/Ceramic Seat, PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/16" Ceramic Ball/Ceramic Seat, PEEK CartridgeWaters 616, AllianceLog in for availabilityEach$655.97Supplier Number: 39-48-01949
- 77101-316OPTI-MAX Outlet Cartridge Check Valve, 1/16" Ceramic Ball/Ceramic Seat, PEEK CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/16" Ceramic Ball/Ceramic Seat, PEEK CartridgeWaters 616, AllianceLog in for availabilityEach$655.97Supplier Number: 39-48-01950
- 77101-322OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, PEEK Cartridge, BiocompatibleOPTI-MAX Inlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, PEEK Cartridge, BiocompatibleWaters 625/626Log in for availabilityEach$653.42Supplier Number: 39-49-01907
- 77101-324OPTI-MAX Outlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, PEEK Cartridge, BiocompatibleOPTI-MAX Outlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, PEEK Cartridge, BiocompatibleWaters 625/626Log in for availabilityEach$653.42Supplier Number: 39-49-01909
- 77101-326OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, PEEK CartridgeWaters 616, AllianceLog in for availabilityEach$655.97Supplier Number: 39-49-01949
- 77101-328OPTI-MAX Outlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, PEEK CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, PEEK CartridgeWaters 616, AllianceLog in for availabilityEach$655.97Supplier Number: 39-49-01950
- 77101-338OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat,SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat,SS CartridgeWaters 100 µL Head 515, 1515, 1525 [M45/45G, M501, 510, 590, 600/600E, 610, 6K/6KA]Log in for availabilityEach$777.11Supplier Number: 39-56-01251
- 77101-340OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS CartridgeWaters 100 µL Head [M45/45G, M501, 510, 590, 600/600E, 610, 6K/6KA] & 225 µL Head (#3) [510, 590, 600/600E, 610, 650, 6K/6KA], Waters 515, 1515, 1525Log in for availabilityEach$786.92Supplier Number: 39-56-01256
- 77101-346OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball & Sapphire Seat, SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball & Sapphire Seat, SS CartridgeWaters 100 µL Head [M45/45G, M501, 510, 590, 600/600E, 610, 6K/6KA, 515, 1515, 1525]Log in for availabilityEach$657.98Supplier Number: 39-57-01251
- 77101-348OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball & Sapphire Seat, SS CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball & Sapphire Seat, SS CartridgeWaters 100 µL Head [M45/45G, M501, 510, 590, 600/600E, 610, 6K/6KA] & 225 µL Head (#3) [510, 590, 600/600E, 610, 650, 6K/6KA, 515, 1515, 1525]Log in for availabilityEach$657.98Supplier Number: 39-57-01256
- 77101-354OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK CartridgeWaters 100 µL Head [M45/45G, M501, 510, 590, 600/600E, 610, 6K/6KA]Log in for availabilityEach$657.98Supplier Number: 39-58-01251
- 77101-356OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK CartridgeWaters 100 µL Head [M45/45G, M501, 510, 590, 600/600E, 610, 6K/6KA] & 225 µL Head (#3) [510, 590, 600/600E, 610, 650, 6K/6KA]Log in for availabilityEach$657.98Supplier Number: 39-58-01256
- 77101-362OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball & Sapphire Seat, PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball & Sapphire Seat, PEEK CartridgeWaters 100 µL Head [M45/45G, M501, 510, 590, 600/600E, 610, 6K/6KA]Log in for availabilityEach$657.98Supplier Number: 39-59-01251
- 77101-364OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball & Sapphire Seat, PEEK CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball & Sapphire Seat, PEEK CartridgeWaters 100 µL Head [M45/45G, M501, 510, 590, 600/600E, 610, 6K/6KA] & 225 µL Head (#3) [510, 590, 600/600E, 610, 650, 6K/6KA]Log in for availabilityEach$657.98Supplier Number: 39-59-01256
- 77101-366OPTI-MAX EF Inlet Cartridge Check Valve 3/16" Ceramic Ball & Seat, SS CartridgeOPTI-MAX EF Inlet Cartridge Check Valve 3/16" Ceramic Ball & Seat, SS CartridgeWaters 225 µL (#3) [510, 590, 600/600E, 610, 650, 6K/6KA]Log in for availabilityEach$778.81Supplier Number: 39-66-01262
- 77101-368OPTI-MAX EF Outlet Cartridge Check Valve 3/16" Ceramic Ball & Seat, SS CartridgeOPTI-MAX EF Outlet Cartridge Check Valve 3/16" Ceramic Ball & Seat, SS CartridgeWaters 400 µL (#4) & 900 µL (#6) Heads [Delta Prep 3000, 600/600E, 650]Log in for availabilityEach$778.81Supplier Number: 39-66-01267
- 77101-370OPTI-MAX EF Inlet Cartridge Check Valve 3/16" Ruby Ball & Sapphire Seat, SS CartridgeOPTI-MAX EF Inlet Cartridge Check Valve 3/16" Ruby Ball & Sapphire Seat, SS CartridgeWaters 225 µL (#3) [510, 590, 600/600E, 610, 650, 6K/6KA]Log in for availabilityEach$773.13Supplier Number: 39-67-01262
- 77101-372OPTI-MAX EF Outlet Cartridge Check Valve 3/16" Ruby Ball & Sapphire Seat, SS CartridgeOPTI-MAX EF Outlet Cartridge Check Valve 3/16" Ruby Ball & Sapphire Seat, SS CartridgeWaters 400 µL (#4) & 900 µL (#6) Heads [Delta Prep 3000, 600/600E, 650]Log in for availabilityEach$773.13Supplier Number: 39-67-01267
- 77101-374OPTI-MAX EF Inlet Cartridge Check Valve 3/16" Ceramic Ball & Seat, PEEK CartridgeOPTI-MAX EF Inlet Cartridge Check Valve 3/16" Ceramic Ball & Seat, PEEK CartridgeWaters 225 µL (#3) [510, 590, 600/600E, 610, 650, 6K/6KA]Log in for availabilityEach$773.13Supplier Number: 39-68-01262
- 77101-376OPTI-MAX EF Outlet Cartridge Check Valve 3/16" Ceramic Ball & Seat, PEEK CartridgeOPTI-MAX EF Outlet Cartridge Check Valve 3/16" Ceramic Ball & Seat, PEEK CartridgeWaters 400 µL (#4) & 900 µL (#6) Heads [Delta Prep 3000, 600/600E, 650]Log in for availabilityEach$773.13Supplier Number: 39-68-01267
- 83008-410Deuterium LampSupplier Number: 39-32-02823
- 83008-412Deuterium LampSupplier Number: 39-32-02825
- 83008-418OPTI-SEAL UHMW-PE Plunger SealOPTI-SEAL UHMW-PE Plunger SealWaters Alliance/2690, 2695, 2790, 2795Log in for availabilityPack of 2$270.32Supplier Number: 39-36-02603
- 83008-454OPTI-MAX Cartridge Check Valve Conversion Kit, 1/8" Ceramic Ball & Seat, SS CartridgeOPTI-MAX Cartridge Check Valve Conversion Kit, 1/8" Ceramic Ball & Seat, SS CartridgeWaters 100 µL/Dual Head SystemsLog in for availabilityEach$3,199.58Supplier Number: 39-56-02166
- 83008-456OPTI-MAX Cartridge Check Valve Conversion Kit, 1/8" Ceramic Ball & Seat, SS CartridgeOPTI-MAX Cartridge Check Valve Conversion Kit, 1/8" Ceramic Ball & Seat, SS CartridgeWaters 100 µL/Single Head SystemsLog in for availabilityEach$1,596.43Supplier Number: 39-56-02186
- 83008-458OPTI-MAX Cartridge Check Valve Conversion KitOPTI-MAX Cartridge Check Valve Conversion KitWaters 225 µL HeadLog in for availabilityEach$3,199.71Supplier Number: 39-66-02167
- 83008-460OPTI-MAX Cartridge Check Valve Conversion KitSupplier Number: 39-46-02168
- 83008-462OPTI-MAX Cartridge Check Valve Conversion KitSupplier Number: 39-46-02187
- 83008-464OPTI-MAX Cartridge Check Valve Conversion KitOPTI-MAX Cartridge Check Valve Conversion KitWaters 400 µL HeadLog in for availabilityEach$3,251.93Supplier Number: 39-66-02169
- 83008-466OPTI-MAX Cartridge Check Valve Conversion KitSupplier Number: 39-48-02170
- 87006-036Face Seals Replacement KitSupplier Number: 39-36-02855
- 87006-038Seal Wash Face Seals Replacement KitSeal Wash Face Seals Replacement KitWaters 2690, 2695, 2790, 2795Log in for availabilityEach$114.20Supplier Number: 39-36-02856
- 87006-042Seal Wash, Plunger Seal, Replacement KitSeal Wash, Plunger Seal, Replacement KitWaters 2690, 2695, 2790, 2795Log in for availabilityEach$233.61Supplier Number: 39-36-02860
Beckman
Product Information
- For use withAvailabilityPrice
- 21509-1873/16" Ball & SeatSupplier Number: 22-38-01311
- 21509-903OPTI-MAX Inlet Cartridge Check Valve, 3/16" Ruby Ball/Sapphire Seat, SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 3/16" Ruby Ball/Sapphire Seat, SS CartridgeAll BeckmanLog in for availabilityEach$700.26Supplier Number: 22-67-00516
- 21509-905OPTI-MAX Inlet Cartridge Check Valve, 3/16" Ceramic Ball & Seat, PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 3/16" Ceramic Ball & Seat, PEEK CartridgeAll BeckmanLog in for availabilityEach$700.26Supplier Number: 22-68-00516
- 21509-906OPTI-MAX Outlet Cartridge Check Valve, 3/16" Ceramic Ball & Seat, PEEK CartridgeOPTI-MAX Outlet Cartridge Check Valve, 3/16" Ceramic Ball & Seat, PEEK CartridgeAll BeckmanLog in for availabilityEach$700.26Supplier Number: 22-68-00520
- 21509-908OPTI-MAX Outlet Cartridge Check Valve, 3/16" Ruby Ball/Sapphire Seat, SS CartridgeOPTI-MAX Outlet Cartridge Check Valve, 3/16" Ruby Ball/Sapphire Seat, SS CartridgeAll BeckmanLog in for availabilityEach$700.26Supplier Number: 22-67-00520
- 77100-540OPTI-MAX Inlet Cartridge Check Valve, 3/16" Ruby Ball/Sapphire Seat, PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 3/16" Ruby Ball/Sapphire Seat, PEEK CartridgeAll BeckmanLog in for availabilityEach$700.26Supplier Number: 22-69-00516
- 77100-542OPTI-MAX Outlet Cartridge Check Valve, 3/16" Ruby Ball/Sapphire Seat, PEEK CartridgeOPTI-MAX Outlet Cartridge Check Valve, 3/16" Ruby Ball/Sapphire Seat, PEEK CartridgeAll BeckmanLog in for availabilityEach$700.26Supplier Number: 22-69-00520
Bischoff
Product Information
- For use withAvailabilityPrice
- 21510-034ITB PTFE Piston Seal, (Microbore Type)ITB PTFE Piston Seal, (Microbore Type)Bischoff 2200, Anspec SM909, Alcott Micromeritics 760Log in for availabilityEach$216.63Supplier Number: 24-36-00573
- 21510-036ITB PTFE Piston Seal, (Microbore Type)ITB PTFE Piston Seal, (Microbore Type)Bischoff 2200, Anspec SM909, Alcott Micromeritics760Log in for availabilityPack of 10$2,875.24Supplier Number: 24-36-00575
- 21510-580Piston Seal Support Ring, (Microbore Type)Piston Seal Support Ring, (Microbore Type)Bischoff 2200, Anspec SM909, Alcott Micromeritics 760Log in for availabilityEach$146.90Supplier Number: 24-36-00574
- 77100-576Piston (Analytical)Piston (Analytical)Bischoff 2200, Anspec SM909, Alcott Micromeritics 760Log in for availabilityEach$643.35Supplier Number: 24-34-00559
- 77100-582OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, SS Cartridge, (Microbore Type)OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, SS Cartridge, (Microbore Type)Bischoff 2200, Anspec SM909, Alcott Micromeritics 760Log in for availabilityEach$660.30Supplier Number: 24-46-00585
- 77100-584OPTI-MAX Outlet Cartridge Check Valve, 1/16 Ceramic Ball & Seat, SS Cartridge, (Microbore Type)OPTI-MAX Outlet Cartridge Check Valve, 1/16 Ceramic Ball & Seat, SS Cartridge, (Microbore Type)Bischoff 2200, Anspec SM909, Alcott Micromeritics 760Log in for availabilityEach$660.30Supplier Number: 24-46-00589
- 77100-586OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, SS Cartridge, (Microbore Type)OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, SS Cartridge, (Microbore Type)Bischoff 2200, Anspec SM909, Alcott Micromeritics 760Log in for availabilityEach$660.30Supplier Number: 24-47-00585
- 77100-590OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, PEEK Cartridge, (Microbore Type)OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, PEEK Cartridge, (Microbore Type)Bischoff 2200, Anspec SM909, Alcott Micromeritics 760Log in for availabilityEach$716.37Supplier Number: 24-48-00585
- 77100-592OPTI-MAX Outlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, PEEK Cartridge, (Microbore Type)OPTI-MAX Outlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, PEEK Cartridge, (Microbore Type)Bischoff 2200, Anspec SM909, Alcott Micromeritics 760Log in for availabilityEach$716.37Supplier Number: 24-48-00589
- 77100-594OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, PEEK Cartridge, (Microbore Type)OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, PEEK Cartridge, (Microbore Type)Bischoff 2200, Anspec SM909, Alcott Micromeritics 760Log in for availabilityEach$716.37Supplier Number: 24-49-00585
- 77100-596OPTI-MAX Outlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, PEEK Cartridge, (Microbore Type)OPTI-MAX Outlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, PEEK Cartridge, (Microbore Type)Bischoff 2200, Anspec SM909, Alcott Micromeritics 760Log in for availabilityEach$716.37Supplier Number: 24-49-00589
- 77100-598OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS Cartridge, (Analytical)OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS Cartridge, (Analytical)Bischoff 2200, Anspec SM909, Alcott Micromeritics 760Log in for availabilityEach$617.08Supplier Number: 24-56-00576
- 77100-600OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS Cartridge, (Analytical)OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS Cartridge, (Analytical)Bischoff 2200, Anspec SM909, Alcott Micromeritics 760Log in for availabilityEach$617.08Supplier Number: 24-56-00581
- 77100-602OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, SS Cartridge, (Analytical)OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, SS Cartridge, (Analytical)Bischoff 2200, Anspec SM909, Alcott Micromeritics 760Log in for availabilityEach$622.91Supplier Number: 24-57-00576
- 77100-604OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, SS Cartridge, (Analytical)OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, SS Cartridge, (Analytical)Bischoff 2200, Anspec SM909, Alcott Micromeritics 760Log in for availabilityEach$622.91Supplier Number: 24-57-00581
- 77100-606OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK Cartridge, (Analytical)OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK Cartridge, (Analytical)Bischoff 2200, Anspec SM909, Alcott Micromeritics 760Log in for availabilityEach$622.91Supplier Number: 24-58-00576
- 77100-608OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK Cartridge, (Analytical)OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK Cartridge, (Analytical)Bischoff 2200, Anspec SM909, Alcott Micromeritics 760Log in for availabilityEach$622.91Supplier Number: 24-58-00581
- 77100-610OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, PEEK Cartridge, (Analytical)OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, PEEK Cartridge, (Analytical)Bischoff 2200, Anspec SM909, Alcott Micromeritics 760Log in for availabilityEach$622.91Supplier Number: 24-59-00576
- 77100-612OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, PEEK Cartridge, (Analytical)OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, PEEK Cartridge, (Analytical)Bischoff 2200, Anspec SM909, Alcott Micromeritics 760Log in for availabilityEach$622.91Supplier Number: 24-59-00581
- 83008-470OPTI-MAX Cartridge Check Valve Conversion KitOPTI-MAX Cartridge Check Valve Conversion KitBischoff 2200, SM909, 760Log in for availabilityEach$1,259.70Supplier Number: 24-56-02140
- 83008-472OPTI-MAX Cartridge Check Valve Conversion KitOPTI-MAX Cartridge Check Valve Conversion KitBischoff 2200 MicroboreLog in for availabilityEach$1,348.01Supplier Number: 24-46-02141
Gilson/Rainin
Product Information
- For use withAvailabilityPrice
- 21509-866OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, PEEK Cartridge, 5, 10 & 25 mL S/SC HeadOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, PEEK Cartridge, 5, 10 & 25 mL S/SC HeadGilson/Rainin HP, HPX, HPXL, SD200/300Log in for availabilityEach$687.11Supplier Number: 26-59-00647
- 21509-872OPTI-SEAL UHMW-PE Plunger Seal 10 mL S/SC HeadOPTI-SEAL UHMW-PE Plunger Seal 10 mL S/SC HeadGilson/Rainin HP, HPX, HPXL, SD200/300Discontinued-Supplier Number: 26-36-00633
- 21509-888OPTI-SEAL UHMW-PE Plunger Seal 5 mL S/SC HeadOPTI-SEAL UHMW-PE Plunger Seal 5 mL S/SC HeadGilson/Rainin HP, HPX, HPXL, SD200/300Log in for availabilityPack of 10$3,533.50Supplier Number: 26-36-00635
- 21510-006ITB PTFE Plunger Seal, 10 mL S/SC HeadITB PTFE Plunger Seal, 10 mL S/SC HeadGilson/Rainin HP, HPX, HPXL, SD200/300Log in for availabilityEach$266.55Supplier Number: 26-36-00630
- 21510-008ITB PTFE Plunger Seal, 10 mL S/SC HeadITB PTFE Plunger Seal, 10 mL S/SC HeadGilson/Rainin HP, HPX, HPXL, SD200/300Log in for availabilityPack of 1$2,658.11Supplier Number: 26-36-00631
- 21510-010OPTI-SEAL UHMW-PE Plunger Seal 10 mL S/SC HeadOPTI-SEAL UHMW-PE Plunger Seal 10 mL S/SC HeadGilson/Rainin HP, HPX, HPXL, SD200/300Discontinued-Supplier Number: 26-36-00632
- 77100-644OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS Cartridge, 5, 10 & 25 mL S/SC HeadOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS Cartridge, 5, 10 & 25 mL S/SC HeadGilson/Rainin HP, HPX, HPXL, SD200/300, ProStar 210, 215Log in for availabilityEach$687.98Supplier Number: 26-56-00638
- 77100-646OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS Cartridge, 5, 10 & 25 mL S/SC HeadOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS Cartridge, 5, 10 & 25 mL S/SC HeadGilson/Rainin HP, HPX, HPXL, SD200/300, ProStar 210, 215Log in for availabilityEach$687.98Supplier Number: 26-56-00647
- 77100-648OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, SS Cartridge, 5, 10 & 25 mL S/SC HeadOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, SS Cartridge, 5, 10 & 25 mL S/SC HeadGilson/Rainin HP, HPX, HPXL, SD200/300Log in for availabilityEach$687.11Supplier Number: 26-57-00638
- 77100-650OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, SS Cartridge, 5, 10 & 25 mL S/SC HeadOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, SS Cartridge, 5, 10 & 25 mL S/SC HeadGilson/Rainin HP, HPX, HPXL, SD200/300Log in for availabilityEach$687.11Supplier Number: 26-57-00647
- 77100-652OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK Cartridge, 5, 10 & 25 mL S/SC HeadOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK Cartridge, 5, 10 & 25 mL S/SC HeadGilson/Rainin HP, HPX, HPXL, SD200/300Log in for availabilityEach$687.11Supplier Number: 26-58-00638
- 77100-654OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK Cartridge, 5, 10 & 25 mL S/SC HeadOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK Cartridge, 5, 10 & 25 mL S/SC HeadGilson/Rainin HP, HPX, HPXL, SD200/300Log in for availabilityEach$687.11Supplier Number: 26-58-00647
- 77100-656OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, PEEK Cartridge, 5, 10 & 25 mL S/SC HeadOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, PEEK Cartridge, 5, 10 & 25 mL S/SC HeadGilson/Rainin HP, HPX, HPXL, SD200/300Log in for availabilityEach$687.11Supplier Number: 26-59-00638
- 83008-478OPTI-MAX Cartridge Check Valve Conversion Kit, 5, 10 & 25 mL HeadOPTI-MAX Cartridge Check Valve Conversion Kit, 5, 10 & 25 mL HeadALL GilsonLog in for availabilityEach$1,402.64Supplier Number: 26-56-02144
Gynotek
Product Information
Hitachi
Product Information
- For use withAvailabilityPrice
- 46600-650OPTI-SEAL UHMW-PE Piston SealOPTI-SEAL UHMW-PE Piston SealHitachi 655, L-6000, L-6200, L-6200A, L-7100 (LaChrom)Log in for availabilityPack of 10$1,701.23Supplier Number: 28-36-00692
- 77100-712OPTI-SEAL UHMW-PE Piston SealOPTI-SEAL UHMW-PE Piston SealHitachi 655, L-6000, L-6200, L-6200A, L-7100 (LaChrom)Log in for availabilityEach$170.48Supplier Number: 28-36-00690
- 77100-714OPTI-SEAL UHMW-PE Piston Seal w/ Hastelloy C SpringOPTI-SEAL UHMW-PE Piston Seal w/ Hastelloy C SpringHitachi 655, L-6000, L-6200, L-6200A, L-7100 (LaChrom)Log in for availabilityEach$186.28Supplier Number: 28-36-00691
- 77100-716OPTI-SEAL UHMW-PE Piston Seal w/ Hastelloy C SpringOPTI-SEAL UHMW-PE Piston Seal w/ Hastelloy C SpringHitachi 655, L-6000, L-6200, L-6200A, L-7100 (LaChrom)Log in for availabilityPack of 10$1,883.44Supplier Number: 28-36-00693
- 77100-722OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, SS CartridgeHitachi L-7100 (LaChrom)Log in for availabilityEach$661.68Supplier Number: 28-46-02092
- 77100-724OPTI-MAX Outlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, SS CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, SS CartridgeHitachi L-7100 (LaChrom)Log in for availabilityEach$661.68Supplier Number: 28-46-02094
- 77100-730OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, SS CartridgeHitachi L-7100 (LaChrom)Log in for availabilityEach$660.39Supplier Number: 28-47-02092
- 77100-732OPTI-MAX Outlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, SS CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, SS CartridgeHitachi L-7100 (LaChrom)Log in for availabilityEach$660.39Supplier Number: 28-47-02094
- 77100-738OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, PEEK CartridgeHitachi L-7100 (LaChrom)Log in for availabilityEach$660.39Supplier Number: 28-48-02092
- 77100-740OPTI-MAX Outlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, PEEK CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, PEEK CartridgeHitachi L-7100 (LaChrom)Log in for availabilityEach$660.39Supplier Number: 28-48-02094
- 77100-746OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, PEEK Cartridge)OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, PEEK Cartridge)Hitachi L-7100 (LaChromLog in for availabilityEach$660.39Supplier Number: 28-49-02092
- 77100-748OPTI-MAX Outlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, PEEK CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, PEEK CartridgeHitachi L-7100 (LaChrom)Log in for availabilityEach$660.39Supplier Number: 28-49-02094
- 83008-438OPTI-MAX Cartridge Check Valve Conversion KitSupplier Number: 28-46-02148
JASC
Product Information
- For use withAvailabilityPrice
- 77101-412Sapphire PistonSapphire PistonJasco BIP-1, 880/980 Series, PU-2080, 2085, 2089, 1580, 1586, 2086, 986Log in for availabilityEach$367.27Supplier Number: 42-34-01354
- 77101-414ITB PTFE Piston SealITB PTFE Piston SealJasco BIP-1, 880/980 Series, PU-2080, 2085, 2089, 1580, 1586, 2086, 986Log in for availabilityEach$267.41Supplier Number: 42-36-01355
- 77101-416ITB PTFE Piston SealITB PTFE Piston SealJasco BIP-1, 880/980 Series, PU-2080, 2085, 2089, 1580, 1586, 2086, 986Log in for availabilityPack of 10$2,587.68Supplier Number: 42-36-01356
- 77101-418OPTI-SEAL UHMW-PE Piston SeaOPTI-SEAL UHMW-PE Piston SeaJasco BIP-1, 880/980 Series, PU-2080, 2085, 2089, 1580, 1586, 2086, 986Log in for availabilityEach$192.14Supplier Number: 42-36-01357
- 77101-420OPTI-SEAL UHMW-PE Piston SealOPTI-SEAL UHMW-PE Piston SealJasco BIP-1, 880/980 Series, PU-2080, 2085, 2089, 1580, 1586, 2086, 986Log in for availabilityPack of 10$1,921.32Supplier Number: 42-36-01358
- 77101-428OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS CartridgeJasco BIP-1Log in for availabilityEach$654.33Supplier Number: 42-56-01038
- 77101-430OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS CartridgeJasco BIP-1Log in for availabilityEach$654.31Supplier Number: 42-56-01043
- 77101-432OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS CartridgeJasco 880/980 Series, 1580 , 2080 , 2086 , 2089Log in for availabilityEach$654.31Supplier Number: 42-56-01544
- 77101-434OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS CartridgeJasco 880/980 Series, 1580 , 2080 , 2086 , 2089Log in for availabilityEach$654.31Supplier Number: 42-56-01546
- 77101-436OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/ Sapphire Seat, SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/ Sapphire Seat, SS CartridgeJasco BIP-1Log in for availabilityEach$654.31Supplier Number: 42-57-01038
- 77101-438OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/ Sapphire Seat, SS CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/ Sapphire Seat, SS CartridgeJasco BIP-1Log in for availabilityEach$654.31Supplier Number: 42-57-01043
- 77101-440OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball & Sapphire Seat, SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball & Sapphire Seat, SS CartridgeJasco 880/980 Series, 1580 , 2080 , 2086 , 2089Log in for availabilityEach$654.31Supplier Number: 42-57-01544
- 77101-442OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball & Sapphire Seat, SS CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball & Sapphire Seat, SS CartridgeJasco 880/980 Series, 1580 , 2080 , 2086 , 2089Log in for availabilityEach$654.31Supplier Number: 42-57-01546
- 77101-444OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK CartridgeJasco BIP-1Log in for availabilityEach$654.31Supplier Number: 42-58-01038
- 77101-446OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK CartridgeJasco BIP-1Log in for availabilityEach$654.31Supplier Number: 42-58-01043
- 77101-448OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK CartridgeJasco 880/980 Series, 1580 , 2080 , 2086 , 2089Log in for availabilityEach$654.31Supplier Number: 42-58-01544
- 77101-450OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK CartridgeJasco 880/980 Series, 1580 , 2080 , 2086 , 2089Log in for availabilityEach$654.31Supplier Number: 42-58-01546
- 77101-452OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/ Sapphire Seat, PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/ Sapphire Seat, PEEK CartridgeJasco BIP-1Log in for availabilityEach$654.31Supplier Number: 42-59-01038
- 77101-454OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/ Sapphire Seat, PEEK CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/ Sapphire Seat, PEEK CartridgeJasco BIP-1Log in for availabilityEach$654.31Supplier Number: 42-59-01043
- 77101-456OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball & Sapphire Seat, PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball & Sapphire Seat, PEEK CartridgeJasco 880/980 Series, 1580 , 2080 , 2086 , 2089Log in for availabilityEach$654.31Supplier Number: 42-59-01544
- 77101-458OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball & Sapphire Seat, PEEK CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball & Sapphire Seat, PEEK CartridgeJasco B1002 880/980 Series, 1580 , 2080 , 2086 , 2089Log in for availabilityEach$654.31Supplier Number: 42-59-01546
- 83008-424Deuterium DoubleLife LampSupplier Number: 42-32-02841
- 83008-426Deuterium DoubleLife LampDeuterium DoubleLife LampJasco UV-1570, -1575, -2075, 975 B/C SeriesDiscontinued-Supplier Number: -
- 83008-484OPTI-MAX Cartridge Check Valve Conversion KitSupplier Number: 42-56-02150
- 83008-486OPTI-MAX Cartridge Check Valve Conversion KitOPTI-MAX Cartridge Check Valve Conversion KitJasco 890/980, 1580 , 2080 , 2086 , 2089Log in for availabilityEach$1,335.73Supplier Number: 42-56-02151
LDC/Milton Roy
Product Information
- For use withAvailabilityPrice
- 21509-248ITB PTFE Plunger Seal, BlackITB PTFE Plunger Seal, BlackLDC/Milton Roy, All Analytical including Constametric, Minimetric, and MinipumpDiscontinued-Supplier Number: -
- 21509-805OPTI-SEAL UHMW-PE Plunger SealOPTI-SEAL UHMW-PE Plunger SealLDC/Milton Roy, All Analytical including Constametric, Minimetric, and MinipumpLog in for availabilityEach$147.23Supplier Number: 31-36-00806
- 77100-780Sapphire PlungerSapphire PlungerLDC/Milton Roy, All Analytical including Constametric, Minimetric, and MinipumpDiscontinued-Supplier Number: 31-34-00738
- 77100-784OPTI-SEAL UHMW-PE Plunger SealOPTI-SEAL UHMW-PE Plunger SealLDC/Milton Roy, All Analytical including Constametric, Minimetric, and MinipumpLog in for availabilityPack of 10$1,474.45Supplier Number: 31-36-00807
- 77100-786ITB PTFE Plunger Seal, BlackITB PTFE Plunger Seal, BlackLDC/Milton Roy, All Analytical including Constametric, Minimetric, and MinipumpDiscontinued-Supplier Number: -
- 77100-788OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS CartridgeLDC/Milton Roy, All Analytical including Constametric, Minimetric, and MinipumpLog in for availabilityEach$654.24Supplier Number: 31-56-00759
- 77100-790OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS CartridgeLDC/Milton Roy, All Analytical including Constametric, Minimetric, and MinipumpLog in for availabilityEach$654.24Supplier Number: 31-56-00764
- 77100-792OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, SS CartridgeLDC/Milton Roy, All Analytical including Constametric, Minimetric, and MinipumpLog in for availabilityEach$654.24Supplier Number: 31-57-00759
- 77100-794OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, SS CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, SS CartridgeLDC/Milton Roy, All Analytical including Constametric, Minimetric, and MinipumpLog in for availabilityEach$654.24Supplier Number: 31-57-00764
- 77100-796OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK CartridgeLDC/Milton Roy, All Analytical including Constametric, Minimetric, and MinipumpLog in for availabilityEach$654.24Supplier Number: 31-58-00759
- 77100-798OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK CartridgeLDC/Milton Roy, All Analytical including Constametric, Minimetric, and MinipumpLog in for availabilityEach$654.24Supplier Number: 31-58-00764
- 77100-800OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, PEEK CartridgeLDC/Milton Roy, All Analytical including Constametric, Minimetric, and MinipumpLog in for availabilityEach$654.24Supplier Number: 31-59-00759
- 77100-802OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, PEEK CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, PEEK CartridgeLDC/Milton Roy, All Analytical including Constametric, Minimetric, and MinipumpLog in for availabilityEach$654.24Supplier Number: 31-59-00764
- 83008-490OPTI-MAX Cartridge Check Valve Conversion KitOPTI-MAX Cartridge Check Valve Conversion KitLDC/Milton Roy Minipump, MinimetricLog in for availabilityEach$1,335.72Supplier Number: 31-56-02152
- 83008-492OPTI-MAX Cartridge Check Valve Conversion KitOPTI-MAX Cartridge Check Valve Conversion KitLDC/Milton Roy Constametric, Mini DuplexLog in for availabilityEach$2,676.97Supplier Number: 31-56-02153
LKB
Product Information
- For use withAvailabilityPrice
- 21510-040ITB PTFE Piston Seal, w/ Back-Up RingSupplier Number: 32-36-01303
- 21510-042ITB PTFE Piston Seal, w/ Back-Up RingsSupplier Number: 32-36-01304
- 77100-808Sapphire PistonSupplier Number: 32-34-00771
- 77100-814OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, SS CartridgeLKB 2150, 2248Log in for availabilityEach$700.34Supplier Number: 32-46-00782
- 77100-816OPTI-MAX Outlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, SS CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, SS CartridgeLKB 2150, 2248Log in for availabilityEach$700.34Supplier Number: 32-46-00787
- 77100-818OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, SS CartridgeLKB 2150, 2248Log in for availabilityEach$700.34Supplier Number: 32-47-00782
- 77100-820OPTI-MAX Outlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, SS CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, SS CartridgeLKB 2150, 2248Log in for availabilityEach$700.34Supplier Number: 32-47-00787
- 77100-822OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, PEEK CartridgeLKB 2150, 2248Log in for availabilityEach$700.34Supplier Number: 32-48-00782
- 77100-824OPTI-MAX Outlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, PEEK CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, PEEK CartridgeLKB 2150, 2248Log in for availabilityEach$700.34Supplier Number: 32-48-00787
- 77100-826OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, PEEK CartridgeLKB 2150, 2248Log in for availabilityEach$700.34Supplier Number: 32-49-00782
- 77100-828OPTI-MAX Outlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, PEEK CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, PEEK CartridgeLKB 2150, 2248Log in for availabilityEach$700.34Supplier Number: 32-49-00787
PERK
Product Information
- For use withAvailabilityPrice
- 21510-438OPTI-SEAL UHMW-PE Piston Seal (Low Pressure)OPTI-SEAL UHMW-PE Piston Seal (Low Pressure)Perkin-Elmer Series 4, 400, 410, 620, Integral 4000Log in for availabilityEach$258.20Supplier Number: 33-36-00804
- 21510-440OPTI-SEAL UHMW-PE Piston Seal (Low Pressure)OPTI-SEAL UHMW-PE Piston Seal (Low Pressure)Perkin-Elmer Series 4, 400, 410, 620, Integral 4000Log in for availabilityPack of 10$2,586.83Supplier Number: 33-36-00805
- 77100-864OPTI-SEAL UHMW-PE Piston SealSupplier Number: 33-36-00806
- 77100-866OPTI-SEAL UHMW-PE Piston SealSupplier Number: 33-36-00807
- 77100-868ITB PFTE Piston SealITB PFTE Piston SealPerkin-Elmer Series 100Discontinued-Supplier Number: -
- 77100-870ITB PTFE Piston SealITB PTFE Piston SealPerkin-Elmer Series 100Discontinued-Supplier Number: -
- 77100-874OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS CartridgePerkin-Elmer Model 250, Series 4, 200, 400, 410, 620, Integral 4000Log in for availabilityEach$693.82Supplier Number: 33-56-00830
- 77100-876OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS CartridgePerkin-Elmer Model 250, Series 4, 200, 400, 410, 620, Integral 4000Log in for availabilityEach$693.82Supplier Number: 33-56-00835
- 77100-882OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball & Sapphire Seat, SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball & Sapphire Seat, SS CartridgePerkin-Elmer Model 250, Series 4, 200, 400, 410, 620, Integral 4000Log in for availabilityEach$692.63Supplier Number: 33-57-00830
- 77100-884OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball & Sapphire Seat, SS CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball & Sapphire Seat, SS CartridgePerkin-Elmer Model 250, Series 4, 200, 400, 410, 620, Integral 4000Log in for availabilityEach$692.63Supplier Number: 33-57-00835
- 77100-890OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK CartridgePerkin-Elmer Model 250, Series 4, 200, 400, 410, 620, Integral 4000Log in for availabilityEach$686.20Supplier Number: 33-58-00830
- 77100-892OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK CartridgePerkin-Elmer Model 250, Series 4, 200, 400, 410, 620, Integral 4000Log in for availabilityEach$692.63Supplier Number: 33-58-00835
- 77100-898OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, PEEK CartridgePerkin-Elmer Model 250, Series 4, 200, 400, 410, 620, Integral 4000Log in for availabilityEach$692.63Supplier Number: 33-59-00830
- 77100-900OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, PEEK CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, PEEK CartridgePerkin-Elmer Model 250, Series 4, 200, 400, 410, 620, Integral 4000Log in for availabilityEach$692.63Supplier Number: 33-59-00835
- 83008-500OPTI-MAX Cartridge Check Valve Conversion KitOPTI-MAX Cartridge Check Valve Conversion KitPerkin-Elmer 4, 200, 400, 410, 4000, Model 250Log in for availabilityEach$2,121.13Supplier Number: 33-56-02157
VARI
Product Information
- For use withAvailabilityPrice
- 21510-410ITB PTFE Piston SealSupplier Number: 38-36-01002
- 21510-412ITB PTFE Piston SealSupplier Number: 38-36-01003
- 21510-422OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, PEEK CartridgeVarian 2010Log in for availabilityEach$699.05Supplier Number: 38-59-01038
- 21510-424OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat,PEEK CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat,PEEK CartridgeVarian 2010Log in for availabilityEach$699.05Supplier Number: 38-59-01043
- 77101-098Sapphire PistonSupplier Number: 38-34-00999
- 77101-102OPTI-SEAL UHMW-PE Piston SealOPTI-SEAL UHMW-PE Piston SealVarian 2010, 2510Discontinued-Supplier Number: 38-36-01000
- 77101-104OPTI-SEAL UHMW-PE Piston SealOPTI-SEAL UHMW-PE Piston SealVarian 2010, 2510Discontinued-Supplier Number: 38-36-01001
- 77101-106ITB PTFE Piston SealITB PTFE Piston SealVarian 9000 Star, 9010, 9012, 9020, 220, 230, 240Log in for availabilityEach$163.97Supplier Number: 38-36-01553
- 77101-108ITB PTFE Piston SealITB PTFE Piston SealVarian 9000 Star, 9010, 9012, 9020, 220, 230, 240Log in for availabilityPack of 10$1,639.92Supplier Number: 38-36-01554
- 77101-110OPTI-SEAL UHMW-PE Piston SealOPTI-SEAL UHMW-PE Piston SealVarian 9000 Star, 9010, 9012 9020, 220, 230, 240Discontinued-Supplier Number: -
- 77101-112OPTI-SEAL UHMW-PE Piston SealOPTI-SEAL UHMW-PE Piston SealVarian 9000 Star, 9010, 9012 9020, 220, 230, 240Discontinued-Supplier Number: -
- 77101-114OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat,SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat,SS CartridgeVarian 2010Log in for availabilityEach$699.05Supplier Number: 38-56-01038
- 77101-116OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat,SS CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat,SS CartridgeVarian 2010Log in for availabilityEach$699.05Supplier Number: 38-56-01043
- 77101-118OPTI-MAX Outlet Cartridge Check Valve 1/8" Ceramic Ball & Seat,SS CartridgeOPTI-MAX Outlet Cartridge Check Valve 1/8" Ceramic Ball & Seat,SS CartridgeVarian 9000 Star, 9010, 9012, 9020, 220, 230, 240Log in for availabilityEach$700.29Supplier Number: 38-56-01067
- 77101-120OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS CartridgeVarian 2510Log in for availabilityEach$699.05Supplier Number: 38-56-01544
- 77101-122OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat,SS CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat,SS CartridgeVarian 2510Log in for availabilityEach$699.05Supplier Number: 38-56-01546
- 77101-124OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, SS CartridgeVarian 2010Log in for availabilityEach$699.05Supplier Number: 38-57-01038
- 77101-126OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, SS CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, SS CartridgeVarian 2010Log in for availabilityEach$699.05Supplier Number: 38-57-01043
- 77101-128OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat,SS CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat,SS CartridgeVarian 9000 Star, 9010Log in for availabilityEach$699.05Supplier Number: 38-57-01067
- 77101-130OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball & Sapphire Seat,SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball & Sapphire Seat,SS CartridgeVarian 2510Log in for availabilityEach$699.05Supplier Number: 38-57-01544
- 77101-132OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball & Sapphire Seat,SS CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball & Sapphire Seat,SS CartridgeVarian 2510Log in for availabilityEach$699.05Supplier Number: 38-57-01546
- 77101-134OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat,PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat,PEEK CartridgeVarian 2010Log in for availabilityEach$699.05Supplier Number: 38-58-01038
- 77101-136OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat,PEEK CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat,PEEK CartridgeVarian 2010Log in for availabilityEach$699.05Supplier Number: 38-58-01043
- 77101-138OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat,PEEK CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat,PEEK CartridgeVarian 9000 Star, 9010Log in for availabilityEach$699.05Supplier Number: 38-58-01067
- 77101-140OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat,PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat,PEEK CartridgeVarian 2510Log in for availabilityEach$699.05Supplier Number: 38-58-01544
- 77101-142OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK CartridgeVarian 2510Log in for availabilityEach$699.05Supplier Number: 38-58-01546
- 77101-144OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat,PEEK CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat,PEEK CartridgeVarian 9000 Star, 9010Log in for availabilityEach$699.05Supplier Number: 38-59-01067
- 77101-146OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball & Sapphire Seat,PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball & Sapphire Seat,PEEK CartridgeVarian 2510Log in for availabilityEach$699.05Supplier Number: 38-59-01544
- 77101-148OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball & Sapphire Seat,PEEK CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball & Sapphire Seat,PEEK CartridgeVarian 2510Log in for availabilityEach$699.05Supplier Number: 38-59-01546
- 83008-452OPTI-MAX Cartridge Check Valve Conversion KitOPTI-MAX Cartridge Check Valve Conversion KitVarian 9000 Star, 9010, 9012Log in for availabilityEach$728.86Supplier Number: 38-56-01704
- 83008-506OPTI-MAX Cartridge Check Valve Conversion KitSupplier Number: 38-56-02164
- 83008-508OPTI-MAX Cartridge Check Valve Conversion KitSupplier Number: 38-56-02165
SHIM
Product Information
- For use withAvailabilityPrice
- 21509-828OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, PEEK CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, PEEK CartridgeShimadzu LC-6A, LC-10ASLog in for availabilityEach$388.17Supplier Number: 34-59-00907
- 21509-826OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, PEEK CartridgeShimadzu LC-6A, LC-10ASLog in for availabilityEach$395.57Supplier Number: 34-59-00902
- 21510-370UHMW-PE/Elastomer Energized Plunger SealUHMW-PE/Elastomer Energized Plunger SealShimadzu LC-600/LC-9A, LC-10AD, LC-10ADVP, LC2010, ESA Model 580, 582, 584Discontinued-Supplier Number: -
- 21510-372UHMW-PE/Elastomer Energized Plunger SealUHMW-PE/Elastomer Energized Plunger SealShimadzu LC-600/LC-9A, LC-10AD, LC-10ADVP, LC2010, ESA Model 580, 582, 584Discontinued-Supplier Number: -
- 21509-962ITB PTFE Plunger SealITB PTFE Plunger SealShimadzu LC-6A, LC-7A, LC-10AS, LC-10AT, LC-10ATVP, LC-20ATLog in for availabilityEach$174.16Supplier Number: 34-36-00863
- 21509-964ITB PTFE Plunger SealITB PTFE Plunger SealShimadzu LC-6A, LC-7A, LC-10AS, LC-10AT, LC-10ATVP, LC-20ATLog in for availabilityPack of 10$1,712.30Supplier Number: 34-36-00864
- 21509-967OPTI-SEAL UHMW-PE Plunger SealOPTI-SEAL UHMW-PE Plunger SealShimadzu LC-10AS, LC-10AT, LC-10ATVP, LC-20AT, LC-6A, LC-7ALog in for availabilityEach$186.23Supplier Number: 34-36-00865
- 77100-924Sapphire PlungerSupplier Number: 34-34-01345
- 77100-930OPTI-SEAL UHMW-PE Plunger SealOPTI-SEAL UHMW-PE Plunger SealShimadzu LC-600/LC-9A, LC-10AD, LC-10ADVP, LC2010, ESA Model 580, 582, 584, LC-20ADXR, LC-20ABXRDiscontinued-Supplier Number: -
- 77100-938OPTI-SEAL UHMW-PE Plunger SealOPTI-SEAL UHMW-PE Plunger SealShimadzu LC-10AS, LC-10AT, LC-10ATVP, LC-20AT, LC-6A, LC-7ALog in for availabilityPack of 10$1,840.10Supplier Number: 34-36-00866
- 77100-946OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, SS CartridgeShimadzu LC-600/LC-9A, LC-10AD, ESA Model 580Log in for availabilityEach$477.55Supplier Number: 34-46-00885
- 77100-948OPTI-MAX Outlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, SS CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, SS CartridgeShimadzu LC-2010, LC-600/LC-9A, LC-10AD/ADVP/AT/ATVPLog in for availabilityEach$477.55Supplier Number: 34-46-00893
- 77100-950OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, SS CartridgeShimadzu LC-10AT/ATVPDiscontinued-Supplier Number: -
- 77100-952OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, SS CartridgeShimadzu LC-600/LC-9A, LC-10AD, ESA Model 580Log in for availabilityEach$486.48Supplier Number: 34-47-00885
- 77100-954OPTI-MAX Outlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, SS CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, SS CartridgeShimadzu LC-2010, LC-600/LC-9A, LC-10AD/ADVP/AT/ATVP, LC2010Log in for availabilityEach$558.38Supplier Number: 34-47-00893
- 77100-956OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, SS CartridgeShimadzu LC-10AT/ATVPDiscontinued-Supplier Number: -
- 77100-958OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, PEEK CartridgeShimadzu LC-600/LC-9A, LC-10AD, ESA Model 580Log in for availabilityEach$486.48Supplier Number: 34-48-00885
- 77100-960OPTI-MAX Outlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, PEEK CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, PEEK CartridgeShimadzu LC-2010, LC-600/LC-9A, LC-10AD/ADVP/AT/ATVP, ESA 582 & 584Log in for availabilityEach$486.48Supplier Number: 34-48-00893
- 77100-962OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, PEEK CartridgeShimadzu LC-10AT/ATVPDiscontinued-Supplier Number: -
- 77100-964OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, PEEK CartridgeShimadzu LC-600/LC-9A, LC-10AD, ESA Model 580Log in for availabilityEach$486.48Supplier Number: 34-49-00885
- 77100-966OPTI-MAX Outlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, PEEK CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, PEEK CartridgeShimadzu LC-2010 LC-600/LC-9A, LC-10AD/ADVP/AT/ATVPLog in for availabilityEach$486.48Supplier Number: 34-49-00893
- 77100-968OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, PEEK CartridgeShimadzu LC-10AT/ATVPDiscontinued-Supplier Number: -
- 77100-970OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS CartridgeShimadzu LC-6A, LC-10ASLog in for availabilityEach$395.57Supplier Number: 34-56-00902
- 77100-972OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, SS CartridgeShimadzu LC-8ALog in for availabilityEach$400.66Supplier Number: 34-56-00907
- 77100-974OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, SS CartridgeShimadzu LC-6A, LC-10ASLog in for availabilityEach$395.57Supplier Number: 34-57-00902
- 77100-976OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, SS CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, SS CartridgeShimadzu LC-6A, LC-10ASLog in for availabilityEach$388.96Supplier Number: 34-57-00907
- 77100-978OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK CartridgeShimadzu LC-6A, LC-10ASLog in for availabilityEach$395.57Supplier Number: 34-58-00902
- 77100-980OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK CartridgeShimadzu LC-6A, LC-10ASLog in for availabilityEach$388.17Supplier Number: 34-58-00907
- 21010-580Sapphire Plunger AssemblySapphire Plunger AssemblyShimadzu LC-10ADVP, LC2010, LC-10AD, LC-600, LC-20ADXR, LC-20ABXRDiscontinued-Supplier Number: -
- 21010-582Sapphire PlungerSupplier Number: 34-34-02650
- 83008-384OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, SS CartridgeShimadzu LC-10ADVP, ESA 582 & 584Log in for availabilityEach$500.81Supplier Number: 34-46-02644
- 83008-386OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, SS CartridgeShimadzu LC-10ADVP, LC2010Log in for availabilityEach$451.66Supplier Number: 34-47-02644
- 83008-388OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/16" Ceramic Ball & Seat, PEEK CartridgeShimadzu LC-10ADVPLog in for availabilityEach$468.71Supplier Number: 34-48-02644
- 83008-390OPTI-MAX Inlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, PEEK CartridgeShimadzu LC-10ADVP, LC-2010Log in for availabilityEach$468.71Supplier Number: 34-49-02644
- 83008-440OPTI-MAX Cartridge Check Valve Conversion KitOPTI-MAX Cartridge Check Valve Conversion KitShimadzu LC-600/9A, LC-10ADLog in for availabilityEach$1,983.41Supplier Number: 34-46-02158
- 83008-442OPTI-MAX Cartridge Check Valve Conversion KitOPTI-MAX Cartridge Check Valve Conversion KitShimadzu LC-6A, LC-10ASLog in for availabilityEach$696.42Supplier Number: 34-56-02159
- 83008-444OPTI-MAX Cartridge Check Valve Conversion KitOPTI-MAX Cartridge Check Valve Conversion KitShimadzu LC-10AT, LC-10ATVPDiscontinued-Supplier Number: -
- 83008-446OPTI-MAX Cartridge Check Valve Conversion KitOPTI-MAX Cartridge Check Valve Conversion KitShimadzu LC-10ADVP, LC-20AD, LC-20AB, LC2010Log in for availabilityEach$1,642.09Supplier Number: 34-46-02627
Spectra Physics
Product Information
- For use withAvailabilityPrice
- 21510-050ITB PTFE Piston SealITB PTFE Piston SealSpectra Physics 8800, 8810, Isochrom, P-SeriesLog in for availabilityEach$168.73Supplier Number: 35-36-00938
- 21510-052ITB PTFE Piston SealITB PTFE Piston SealSpectra Physics 8800, 8810, Isochrom, P-SeriesLog in for availabilityPack of 10$1,684.11Supplier Number: 35-36-00939
- 77100-998Deuterium LampDeuterium LampSpectra Physics Focus, 100, 200, UV-100, -150, -200, -100, 2000, 3000, Focus, CE SeriesLog in for availabilityEach$2,690.48Supplier Number: 35-32-01333
- 77101-002PistonPistonSpectra Physics/TSP 8800, 8810, Isochrom, P-SeriesDiscontinued-Supplier Number: -
- 77101-004Back-Up Seal, PCTFESupplier Number: 35-36-00916
- 77101-006Back-Up Seal, PCTFESupplier Number: 35-36-00917
- 77101-008OPTI-SEAL UHMW-PE Piston SealOPTI-SEAL UHMW-PE Piston SealSpectra Physics/TSP 8800 Flush Seal, 8810, IsoChrom, P-SeriesDiscontinued-Supplier Number: -
- 77101-010OPTI-SEAL UHMW-PE Piston SealOPTI-SEAL UHMW-PE Piston SealSpectra Physics/TSP 8800 Flush Seal, 8810, IsoChrom, P-SeriesDiscontinued-Supplier Number: -
- 77101-012ITB PTFE Flush SealITB PTFE Flush SealSpectra Physics/TSP 8800, 8810, IsoChrom, P-SeriesLog in for availabilityEach$145.23Supplier Number: 35-36-00920
- 77101-014ITB PTFE Flush SealITB PTFE Flush SealSpectra Physics/TSP 8800, 8810, IsoChrom, P-SeriesLog in for availabilityPack of 10$1,452.41Supplier Number: 35-36-00921
- 77101-016OPTI-SEAL UHMW-PE Piston SealOPTI-SEAL UHMW-PE Piston SealSpectra Physics 8800, 8810, Isochrom, P-SeriesLog in for availabilityEach$158.99Supplier Number: 35-36-00930
- 77101-018OPTI-SEAL UHMW-PE Piston SealOPTI-SEAL UHMW-PE Piston SealSpectra Physics 8800, 8810, Isochrom, P-SeriesLog in for availabilityPack of 10$1,589.91Supplier Number: 35-36-00931
- 77101-026OPTI-MAX Transducer Cartridge Check Valve, 1/8" Ceramic Ball & Seat,SS CartridgeOPTI-MAX Transducer Cartridge Check Valve, 1/8" Ceramic Ball & Seat,SS CartridgeSpectra Physics P-SeriesLog in for availabilityEach$852.16Supplier Number: 35-56-00952
- 77101-028OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat,SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat,SS CartridgeSpectra Physics 8700, 8800, 8810, IsoChrom, P-SeriesLog in for availabilityEach$852.16Supplier Number: 35-56-00956
- 77101-030OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat,SS CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat,SS CartridgeSpectra Physics 8700, 8800, 8810, IsoChromLog in for availabilityEach$853.57Supplier Number: 35-56-00961
- 77101-032OPTI-MAX Transducer Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, SS CartridgeOPTI-MAX Transducer Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, SS CartridgeSpectra Physics P-SeriesLog in for availabilityEach$852.16Supplier Number: 35-57-00952
- 77101-034OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat,SS CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat,SS CartridgeSpectra Physics 8700, 8800, 8810, IsoChrom, P-SeriesLog in for availabilityEach$852.16Supplier Number: 35-57-00956
- 77101-036OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, SS CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, SS CartridgeSpectra Physics 8700, 8800, 8810, IsoChromLog in for availabilityEach$852.16Supplier Number: 35-57-00961
- 77101-038OPTI-MAX Transducer Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK CartridgeOPTI-MAX Transducer Cartridge Check Valve, 1/8" Ceramic Ball & Seat, PEEK CartridgeSpectra Physics P-SeriesLog in for availabilityEach$852.16Supplier Number: 35-58-00952
- 77101-040OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat,PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat,PEEK CartridgeSpectra Physics 8700, 8800, 8810, IsoChrom, P-SeriesLog in for availabilityEach$852.16Supplier Number: 35-58-00956
- 77101-042OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat,PEEK CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ceramic Ball & Seat,PEEK CartridgeSpectra Physics 8700, 8800, 8810, IsoChromLog in for availabilityEach$852.16Supplier Number: 35-58-00961
- 77101-044OPTI-MAX Transducer Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, PEEK CartridgeOPTI-MAX Transducer Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, PEEK CartridgeSpectra Physics P-SeriesLog in for availabilityEach$852.16Supplier Number: 35-59-00952
- 77101-046OPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, PEEK CartridgeOPTI-MAX Inlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat, PEEK CartridgeSpectra Physics 8700, 8800, 8810, IsoChrom, P-SeriesLog in for availabilityEach$852.16Supplier Number: 35-59-00956
- 77101-048OPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat,PEEK CartridgeOPTI-MAX Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire Seat,PEEK CartridgeSpectra Physics 8700, 8800, 8810, IsoChromLog in for availabilityEach$852.16Supplier Number: 35-59-00961
- 83008-448OPTI-MAX Cartridge Check Valve Conversion KitOPTI-MAX Cartridge Check Valve Conversion KitSpectra Physics 8700, 8800, 8810, IsochromLog in for availabilityEach$1,471.27Supplier Number: 35-56-02161
- 83008-450OPTI-MAX Cartridge Check Valve Conversion KitOPTI-MAX Cartridge Check Valve Conversion KitSpectra Physics P-SeriesLog in for availabilityEach$1,471.27Supplier Number: 35-56-02162
Accessories
Product Information
- Catalog #AvailabilityPrice
- Optimize Seal Forming/Insertion Kit, Small/Medium/LargeOptimize Seal Forming/Insertion Kit, Small/Medium/Large83008-150Log in for availabilityPack of 3 - $344.08
Product Information
- Catalog #AvailabilityPrice
- Optimize Seal Forming/Insertion Kit, Small
Product Information
- Catalog #AvailabilityPrice
- Optimize Seal Forming/Insertion Kit, Medium
Product Information
- Catalog #AvailabilityPrice
- Optimize Seal Forming/Insertion Kit, Large