ContactUsLinkComponent
MCl-186, Calbiochem
  80056-994
 :  
MCl-186, Calbiochem
  80056-994
 :  443300-500MG
 :  
  • Size:
    500 mg
  • Cat. No.:
    80056-994
  • Supplier no.:
    443300-500MG
A free radical scavenger and antioxidant that reduces post-ischemic brain injury
Inhibits iron-dependent peroxidation in rat brain homogenates (IC50=15µM) and mitochondrial homogenates (IC50=2.3µM).