ContactUsLinkComponent
Protective Eyewear Slip-On Side Shields, 3M™
Protective Eyewear Slip-On Side Shields, 3M™
Catalog # 33001-569
Supplier:  Avantor
undefined
Protective Eyewear Slip-On Side Shields, 3M™
Catalog # 33001-569
Supplier:  Avantor
Specifications
  • Description:
    Protective eyewear slip-on side shields
  • Environmentally Preferable:
  • Cat. No.:
    33001-569
  • Lens tint:
    Clear
  • Supplier no.:
    23451-00030-20
Specifications
About this item
Protective eyewear slip-on side shields.
Shields can be used with a variety of different eyewear styles.
Caution: Note: Side shields do not meet ANSI standards.